Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This monoclonal antibody can be used to detect a sub-set of the non-snRNP splicing factors termed SR proteins. The antibody detects SRp75, SRp55, SRp40, and SRp20 but not SRp30a or b (ASF/SF2, SC-35) proteins. 16H3 also recognizes approximately 20 distinct nuclear proteins including the U1 70K component of the U1 snRNP and both subunits of U2AF. The epitope detected by the 16H3 monoclonal antibody has been termed the 'alternating arginine domain' and is composed almost exclusively of argnine alternating with glutamate and aspartate. 16H3 targets are predominantly nuclear, nonnucleolar, and are localized to active sites of polymerase II transcription.
SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Delayed-early protein HRS; epididymis secretory protein Li 91; epididymis secretory sperm binding protein; Insulin-induced growth response protein CL-4; pre-mRNA splicing factor SRp20; Pre-mRNA-splicing factor SRP20; Pre-mRNA-splicing factor SRP40; Pre-mRNA-splicing factor SRP55; Pre-mRNA-splicing factor SRP75; Protein X16; RNA binding protein; Serine/arginine-rich splicing factor 3; Serine/arginine-rich splicing factor 4; Serine/arginine-rich splicing factor 5; Serine/arginine-rich splicing factor 6; SFRS5; splicing factor arginine/serine-rich 5 (SRp40 HRS); splicing factor SRp55; splicing factor SRp55-1; Splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); Splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); Splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); Splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB; unnamed protein product
Gene Aliases: 1210001E11Rik; 5730499P16Rik; AI314910; AL024116; AW146126; AW550192; B52; Cl-4; HEL-S-91; HRS; MNCb-2616; RGD1561347; SFRS3; SFRS4; SFRS5; SFRS6; SRP20; SRP40; SRP55; SRP55-1; SRP75; SRSF3; SRSF4; SRSF5; SRSF6; X16
UniProt ID: (Bovine) Q3SZR8, (Human) P84103, (Human) Q08170, (Human) Q13243, (Human) Q13247, (Bovine) Q3B7L6, (Mouse) P84104, (Mouse) O35326, (Rat) Q09167, (Rat) G3V6S8, (Mouse) Q3TWW8
Entrez Gene ID: (Dog) 403687, (Chicken) 419815, (Bovine) 540276, (Human) 6428, (Rabbit) 100347748, (Rabbit) 100350179, (Bovine) 614310, (Chicken) 419600, (Dog) 478160, (Human) 6429, (Rabbit) 100340192, (Dog) 490754, (Chicken) 423258, (Human) 6430, (Bovine) 510474, (Human) 6431, (Chicken) 419116, (Bovine) 507828, (Dog) 403667, (Rabbit) 100008654, (Mouse) 20383, (Rat) 361814, (Rat) 362612, (Mouse) 57317, (Mouse) 20384, (Rat) 29667, (Rat) 362264, (Mouse) 67996
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support