Search
Search
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
The protein encoded by this gene is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The encoded nuclear protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in this gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DLDBP; E3BP; Glucose-sensitive factor; GSF; IDX-1; Insulin promoter factor 1; insulin promoter factor 1, homeodomain transcription factor; Insulin upstream factor 1; IPF-1; Islet/duodenum homeobox-1; IUF-1; Pancreas/duodenum homeobox protein 1; pancreatic-duodenal homeobox factor 1; PDHX; PDX-1; somatostatin transcription factor 1; Somatostatin-transactivating factor 1; STF-1
Gene Aliases: GSF; IDX-1; IPF1; IUF1; MODY4; PAGEN1; PDX-1; PDX1; STF-1; STF1
UniProt ID: (Human) P52945
Entrez Gene ID: (Human) 3651
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support