Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Connexin 36 Antibodies

          Invitrogen

          Connexin 36 Monoclonal Antibody (1E5H5)

          Advanced Verification
          48 Published Figures
          59 References
          View all (13) Connexin 36 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Connexin 36 Monoclonal Antibody (1E5H5)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (48)
          • Advanced Verification (1)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 50

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Connexin 36 Antibody (37-4600) in IHC

          Immunofluorescence was performed on sections of adult mouse retina. Tissue sections were probed overnight at 4°C with mouse monoclonal anti-Cx36 antibody (Product # 37-4600, 2 µg/mL), and detected with donkey anti-mouse Alexa Fluor® 488 conjugate. Images were taken on a Zeiss Z2 scanning microscope at 40X magnification, and demonstrate antibody labeling for Cx36 localized at gap junctions between neurons in... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunohistochemistry (Paraffin), Immunocytochemistry (IHC (P), ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunocytochemistry (ICC/IF)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (IHC)
          Connexin 36 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Connexin 36 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Western Blot (WB)
          Connexin 36 Antibody in Immunoprecipitation (IP)
          Connexin 36 Antibody in Immunoprecipitation (IP)
          Connexin 36 Antibody in Immunoprecipitation (IP)
          Connexin 36 Antibody in Immunoprecipitation (IP)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody in Immunomicroscopy (IM)
          Connexin 36 Antibody
          Connexin 36 Monoclonal Antibody (1E5H5)

          Product Details

          37-4600

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          2-3 µg/mL
          View 21 publications 21 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          2-3 µg/mL
          View 10 publications 10 publications

          Immunoprecipitation (IP)

          -
          View 5 publications 5 publications

          Immunomicroscopy (IM)

          -
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Bat, Fish, Human, Mouse, Rat, Sheep

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1E5H5

          Immunogen

          Synthetic peptide derived from the C-terminal region of the rat and mouse Connexin36 proteins.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533320

          Target Information

          Connexin-36 (CX36), is a member of the connexin gene family that is expressed predominantly in mammalian neurons. Connexins associate in groups of 6 and are organized radially around a central pore to form connexons. Each gap junction intercellular channel is formed by the conjunction of 2 connexons. Mutations in connexin-encoding genes can lead to functional and developmental abnormalities.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Connexin-36; Cx36; Gap junction alpha-9 protein; Gap junction delta-2 protein; gap junction membrane channel protein alpha 9; gap junction protein, alpha 9; gap junction protein, delta 2, 36kDa; GJA10

          View more View less

          Gene Aliases: connexin36; CX36; GJA9; GJD2

          View more View less

          UniProt ID: (Human) Q9UKL4, (Mouse) O54851

          View more View less

          Entrez Gene ID: (Human) 57369, (Mouse) 14617

          View more View less

          Function(s)
          gap junction channel activity protein binding gap junction
          Process(es)
          cell communication cell-cell signaling chemical synaptic transmission visual perception neuronal action potential transmembrane transport action potential
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline