Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CYLD Antibodies

          Invitrogen

          CYLD Monoclonal Antibody (733)

          Advanced Verification
          2 Published Figures
          3 References
          View all (19) CYLD antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CYLD Monoclonal Antibody (733)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (2)
          • Advanced Verification (1)
          CYLD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CYLD Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 8

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CYLD Antibody (43-7700) in ICC/IF

          Immunofluorescence analysis of CYLD was done on 70% confluent log phase SHSY5Y cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with CYLD Mouse Monoclonal Antibody (Product # 43-7700) at 1 µg/mL in 1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # A28175) at a dilution... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CYLD Antibody in Immunocytochemistry (ICC/IF)
          CYLD Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CYLD Antibody in Western Blot (WB)
          CYLD Antibody in Western Blot (WB)
          CYLD Antibody in Western Blot (WB)
          CYLD Antibody in Western Blot (WB)
          CYLD Antibody in Western Blot (WB)
          CYLD Antibody
          CYLD Monoclonal Antibody (733)

          Product Details

          43-7700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:50
          -

          Immunocytochemistry (ICC/IF)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Horse, Human, Mouse, Non-human primate, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          733

          Immunogen

          Recombinant protein derived from the N-terminus of human CYLD protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533539

          Target Information

          CYLD is a 956 aa, cytoplasmic, deubiquitinating enzyme belonging to the ubiquitin carboxy-terminal hydrolases (UCH) family of proteins with three cytoskeletal-associated protein-glycine-conserved (CAP-GLY) domains, a proline rich region, a SH3 binding domain and a sequence homology to catalytic domain of UCH. CYLD is identified as a tumor suppressor protein affecting the JNK signaling pathway. CYLD is a negative regulator of TRAF2 and NF-kappa-B signaling pathway and is also known to have receptor-dependent role in regulating the I-kappa-B kinase pathway. It also has a deubiquitinating activity that is directed towards non-Lys-48-linked polyubiquitin chains and TRAP1 is a novel substrate for deubiquitination. Mutated CYLD is known to be associated with cylindromatosis, multiple familial trichoepithelioma, and Brooke-Spiegler syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cyld; cylindromatosis (turban tumor syndrome); Deubiquitinating enzyme CYLD; FLJ20180; FLJ31664; FLJ78684; probable ubiquitin carboxyl-terminal hydrolase CYLD; retinitis pigmentosa 1 homolog; Ubiquitin carboxyl-terminal hydrolase CYLD; ubiquitin specific peptidase like 2; Ubiquitin thioesterase CYLD; ubiquitin thiolesterase CYLD; Ubiquitin-specific-processing protease CYLD; unnamed protein product

          View more View less

          Gene Aliases: 2010013M14Rik; 2900009M21Rik; BRSS; C130039D01Rik; CDMT; CYLD; CYLD1; CYLDI; EAC; FTDALS8; HSPC057; KIAA0849; LRRGT00003; MFT; MFT1; mKIAA0849; Rp1; Rp1h; SBS; TEM; USPL2

          View more View less

          UniProt ID: (Bovine) Q1RMU2, (Human) Q9NQC7, (Rat) Q66H62, (Mouse) Q80TQ2

          View more View less

          Entrez Gene ID: (Horse) 100050308, (Bovine) 536421, (Human) 1540, (Rat) 312937, (Mouse) 74256

          View more View less

          Function(s)
          thiol-dependent ubiquitin-specific protease activity zinc ion binding protein kinase binding Lys63-specific deubiquitinase activity proline-rich region binding Lys48-specific deubiquitinase activity protein binding peptidase activity cysteine-type peptidase activity hydrolase activity metal ion binding ubiquitinyl hydrolase activity, acting on linear ubiquitin molecular_function structural constituent of ribosome thiol-dependent ubiquitinyl hydrolase activity ribosomal protein
          Process(es)
          ubiquitin-dependent protein catabolic process regulation of tumor necrosis factor-mediated signaling pathway Wnt signaling pathway protein deubiquitination negative regulation of NF-kappaB transcription factor activity CD4-positive or CD8-positive, alpha-beta T cell lineage commitment regulation of protein binding innate immune response regulation of B cell differentiation positive regulation of T cell differentiation negative regulation of JNK cascade homeostasis of number of cells regulation of inflammatory response positive regulation of T cell receptor signaling pathway regulation of necroptotic process regulation of microtubule cytoskeleton organization protein K63-linked deubiquitination protein K48-linked deubiquitination negative regulation of canonical Wnt signaling pathway ripoptosome assembly involved in necroptotic process negative regulation of NIK/NF-kappaB signaling regulation of cilium assembly negative regulation of p38MAPK cascade positive regulation of cellular protein localization protein linear deubiquitination immune system process proteolysis I-kappaB kinase/NF-kappaB signaling regulation of mitotic cell cycle negative regulation of type I interferon production nucleotide-binding domain, leucine rich repeat containing receptor signaling pathway defense response to bacterium positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of inflammatory response necroptotic process negative regulation of interleukin-18-mediated signaling pathway positive regulation of extrinsic apoptotic signaling pathway regulation of intrinsic apoptotic signaling pathway cytoplasmic translation biological_process negative regulation of NF-kappaB import into nucleus negative regulation of T cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline