Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • eIF2b epsilon Antibodies

          Invitrogen

          Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody

          Advanced Verification
          3 Published Figures
          7 References
          View all (17) eIF2b epsilon antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (3)
          • Advanced Verification (1)
          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-eIF2b epsilon (Ser539) Antibody (44-530G) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of L6 (Lane 1), L6 differentiated to myotubes (Lane 2), L6 treated with Insulin (10nM for 10 min) (Lane 3), C2C12 (Lane 4) and C2C12 treated with Insulin (10nM for 10 min) ( (Lane 5). The blot was probed with Anti-Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody (Product # 44-530G, 1:2000 dilution) and detected by chemiluminescence ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Phospho-eIF2b epsilon (Ser539) Antibody in Western Blot (WB)
          Phospho-eIF2b epsilon (Ser539) Antibody
          Phospho-eIF2b epsilon (Ser539) Polyclonal Antibody

          Product Details

          44-530G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 7 publications 7 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat, Tag

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of rat eIF2Be that contains serine 535 (corresponding to serine 539 in the human sequence).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533675

          Target Information

          This gene encodes one of five subunits of eukaryotic translation initiation factor 2B, a GTP exchange factor for eukaryotic initiation factor 2 and an essential regulator for protein synthesis. Mutations in this gene and the genes encoding other EIF2B subunits have been associated with leukoencephalopathy with vanishing white matter.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: eIF-2B GDP-GTP exchange factor subunit epsilon; eIF2B GDP-GTP exchange factor subunit epsilon; eukaryotic translation initiation factor 2B, subunit 5 epsilon, 82kDa; initiation factor eIF-2Be; translation initiation factor eIF-2B subunit epsilon; Translation initiation factor eIF2B subunit epsilon; unnamed protein product

          View more View less

          Gene Aliases: C81315; CACH; CLE; EIF-2B; EIF2B5; EIF2BE; EIF2Bepsilon; LVWM; VWM5

          View more View less

          UniProt ID: (Human) Q13144, (Mouse) Q8CHW4, (Rat) Q64350

          View more View less

          Entrez Gene ID: (Human) 8893, (Mouse) 224045, (Rat) 192234

          View more View less

          Function(s)
          translation initiation factor activity guanyl-nucleotide exchange factor activity protein binding translation initiation factor binding translation initiation factor
          Process(es)
          ovarian follicle development cytoplasmic translational initiation translation translational initiation response to heat response to glucose astrocyte development oligodendrocyte development hippocampus development response to endoplasmic reticulum stress myelination positive regulation of apoptotic process response to peptide hormone positive regulation of translation positive regulation of translational initiation astrocyte differentiation T cell receptor signaling pathway regulation of translation negative regulation of translational initiation in response to stress cellular response to drug positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline