Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FAK Antibodies

          Invitrogen

          Phospho-FAK (Tyr397) Monoclonal Antibody (141-9)

          44 Published Figures
          62 References
          View all (108) FAK antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospho-FAK (Tyr397) Monoclonal Antibody (141-9)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (44)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 48

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-FAK (Tyr397) Antibody (44-625G) in ICC/IF

          HeLa cells were fixed prior to immunostaining with the Rabbit anti-FAK (pY397) monoclonal antibody. The signal was detected with an anti-rabbit FITC conjugated secondary antibody. The data shows that the antibody detected phosphorylated FAK localized at focal adhesions. Cells were counterstained with DAPI. (Product # 44-625G) {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FAK (Tyr397) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-FAK (Tyr397) Antibody in Immunohistochemistry (IHC)
          Phospho-FAK (Tyr397) Antibody in Immunohistochemistry (IHC)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Antibody in Western Blot (WB)
          Phospho-FAK (Tyr397) Monoclonal Antibody (141-9)

          Product Details

          44-625G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 39 publications 39 publications

          Immunohistochemistry (IHC)

          -
          View 10 publications 10 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 10 publications 10 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Chicken, Fruit fly, Human, Mouse, Rat, Xenopus

          Published species

          Fish, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          141-9

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human FAK that contains tyrosine 397. The sequence is conserved in mouse, rat, chicken and frog.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 50% glycerol, 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533702

          Product Specific Information

          This antibody will cross-react with the corresponding autophosphorylation site on Proline-rich/Ca2+- activated tyrosine kinase (Pyk2), [pY402]. FAK [pY397] polyclonal antibody (Cat. No. 44-624G) does not cross-react with Pyk2.

          The antibody has been used in western blotting. Previous lots of this antibody have been used in immunocytochemistry. For western blotting applications, we recommend using the antibody at a 1:1,000 starting dilution. For immunofluorecence staining applications, we recommend a 1:50 starting dilution. Positive control used: Primary chicken embryo fibroblasts expressing human FAK and plated on fibronectin.

          We recommend a brief centrifugation before opening to settle vial contents. Then, apportion into working aliquots and store at -20° C. For shipment or short-term storage (up to one week), 2-8° C is sufficient. Expires one year from date of receipt when stored as instructed.

          Target Information

          Focal Adhesion Kinase (FAK) is a 125 kDa non-receptor protein tyrosine kinase that acts as a substrate for Src and is a key element of integrin signaling. FAK plays an important role in cell spreading, differentiation, migration, cell death, and acceleration of the G1 to S phase transition of the cell cycle. FAK has a central catalytic domain and a C-terminal tail that localizes it to focal adhesions, which are sites where cells attach to the extracellular matrix via surface integrin receptors. Increased FAK tyrosine phosphorylation occurs upon integrin engagement with fibronectin. Adhesion of murine NIH3T3 fibroblasts to fibronectin promotes association of the Grb2 adapter protein and c-Src PTK with FAK in vivo, and also results in activation of the ERK2 MAP kinase. In v-Src-transformed NIH3T3, the association of v-Src, Grb2, and Sos with FAK is independent of cell adhesion to fibronectin. In vitro the Grb2 SH2 domain binds directly to tyrosine-phosphorylated FAK, and the binding site has been identified as Tyr925 by site directed mutagenesis. Several transcript variants encoding different isoforms have been found for the FAK gene, but the full-length natures of only three of them have been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CG10023-PA; CG10023-PB; CG10023-PC; CG10023-PD; CG10023-PF; CG10023-PG; EC 2.7.10.2; FADK 1; FAK-Del33; exon 33 deletion splicing mutant; Fak-PA; Fak-PB; Fak-PC; Fak-PD; Fak-PF; Fak-PG; FAK-related non-kinase polypeptide; Focal adhesion kinase 1; focal adhesion kinase homolog; focal adhesion kinase pp125FAK; Focal adhesion kinase-related nonkinase; focal ashension kinase 1; FRNK; p125FAK; p41/p43FRNK; pp125 PTK2; pp125FAK; PPP1R71; Protein phosphatase 1 regulatory subunit 71; protein tyrosine kinase 2; Protein-tyrosine kinase 2; PTK2 protein tyrosine kinase 2; tyrosine kinase; unnamed protein product

          View more View less

          Gene Aliases: CG10023; CT28129; DFAK; DFak56; Dmel\CG10023; Dmel_CG10023; DmFAK; FADK; FADK 1; FAK; FAK1; Fak56; Fak56D; FAK65D; FRNK; Kiaa4203; mKIAA4203; p125FAK; pFAK; pp125FAK; PPP1R71; PTK2; XELAEV_18032408mg; XPFAK

          View more View less

          UniProt ID: (Human) Q05397, (Mouse) P34152, (Rat) O35346, (Xenopus) Q91738

          View more View less

          Entrez Gene ID: (Fruit fly) 37233, (Human) 5747, (Chicken) 396416, (Mouse) 14083, (Rat) 25614, (Xenopus) 399286

          View more View less

          Function(s)
          non-membrane spanning protein tyrosine kinase activity ATP binding nucleotide binding actin binding protein kinase activity protein tyrosine kinase activity protein tyrosine phosphatase activity integrin binding protein binding JUN kinase binding kinase activity transferase activity protein kinase binding protein phosphatase binding SH2 domain binding molecular function activator activity protease binding calcium-dependent cysteine-type endopeptidase activity receptor binding identical protein binding signal transducer activity transferase activity, transferring phosphorus-containing groups protein complex binding phosphatidylinositol 3-kinase binding
          Process(es)
          MAPK cascade microtubule cytoskeleton organization angiogenesis blood vessel development vasculogenesis neuron migration regulation of protein phosphorylation positive regulation of protein phosphorylation protein phosphorylation movement of cell or subcellular component cell adhesion signal complex assembly epidermal growth factor receptor signaling pathway transforming growth factor beta receptor signaling pathway integrin-mediated signaling pathway JNK cascade multicellular organism development positive regulation of cell proliferation regulation of cell shape regulation of epithelial cell migration positive regulation of phosphatidylinositol 3-kinase signaling positive regulation of smooth muscle cell migration phosphorylation peptidyl-tyrosine phosphorylation central nervous system neuron axonogenesis negative regulation of cell-cell adhesion regulation of cell adhesion extracellular matrix organization positive regulation of cell growth positive regulation of cell migration negative regulation of cell migration cellular chloride ion homeostasis regulation of cell adhesion mediated by integrin peptidyl-tyrosine autophosphorylation establishment of nucleus localization regulation of cell proliferation negative regulation of apoptotic process endothelial cell migration innate immune response regulation of osteoblast differentiation positive regulation of cell adhesion positive regulation of protein kinase activity positive regulation of vasodilation negative regulation of organ growth protein autophosphorylation ephrin receptor signaling pathway positive regulation of smooth muscle cell proliferation positive regulation of phagocytosis negative regulation of axonogenesis positive regulation of synaptic transmission regulation of focal adhesion assembly positive regulation of protein kinase B signaling negative regulation of synapse assembly positive regulation of glial cell proliferation growth hormone receptor signaling pathway cellular response to transforming growth factor beta stimulus regulation of substrate adhesion-dependent cell spreading positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of anoikis placenta development regulation of cell-matrix adhesion heart morphogenesis axon guidance regulation of endothelial cell migration positive regulation of epithelial cell migration positive regulation of epithelial to mesenchymal transition positive regulation of macrophage chemotaxis positive regulation of fibroblast migration cell migration establishment of cell polarity cell differentiation detection of muscle stretch netrin-activated signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis regulation of GTPase activity regulation of cell differentiation vascular endothelial growth factor receptor signaling pathway cell motility regulation of multicellular organismal process regulation of cytoskeleton organization positive regulation of wound healing vascular endothelial cell response to oscillatory fluid shear stress positive regulation of macrophage proliferation cytoskeleton organization response to mechanical stimulus response to glucose response to organic substance response to organonitrogen compound response to organic cyclic compound response to estradiol response to drug fat cell differentiation response to arsenic-containing substance proteolysis actin filament organization transmembrane receptor protein tyrosine kinase signaling pathway response to pH negative regulation of cell-substrate adhesion pyramidal neuron migration negative regulation of protein autophosphorylation positive regulation of protein binding response to muscle stretch wound healing, spreading of cells positive regulation of focal adhesion assembly angiogenesis involved in wound healing positive regulation of protein tyrosine kinase activity positive regulation of substrate-dependent cell migration, cell attachment to substrate negative regulation of receptor recycling negative regulation of synaptic growth at neuromuscular junction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline