Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 16
45-6100 was used successfully in the western blot analysis of cytochrome C using human breast cancer cell line lysates.
Positive control: Isolated mitochondria from Human, Bovine, Rat and Mouse heart; Human fibroblasts; HeLa cells.
WB: Predicted molecular weight: 14 kDa. ICC/IF: Heat induced antigen retrieval in 0.1 M Tris-HCl, 5% urea, pH 9.5 for 5 min at 95°C improves signal.
Cytochrome C is an electron transporting protein that resides within the intermembrane space of the mitochondria, where it plays a critical role in oxidative phosphorylation and the production of cellular ATP. Release of cytochrome C from the mitochondria to the cytosol is triggered by apoptotic stimuli. In the cytoplasm, the cytochrome C binds apoptotic protease activating factor which activates the apoptotic initiator procaspase 9. Cytosolic cytochrome C functions in the activation of caspase 3, an ICE family molecule that is a key effector of apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRF; CYCS; Cytochrome c; Cytochrome C, expressed in somatic tissues; Cytochrome c, somatic; hc1; unnamed protein product
Gene Aliases: CYC; CYCS; CYCSA; cytc; HCS; THC4
UniProt ID: (Human) P99999, (Bovine) P62894, (Mouse) P62897, (Rat) P62898
Entrez Gene ID: (Human) 54205, (Bovine) 510767, (Mouse) 13063, (Rat) 25309
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support