Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD8 Antibodies

          Invitrogen

          CD8a Monoclonal Antibody (53-6.7), eFluor™ 450, eBioscience™

          97 Published Figures
          290 References
          View all (628) CD8 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD8a Monoclonal Antibody (53-6.7), eFluor™ 450, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (97)
          CD8a Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD8a Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 98

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD8a Antibody (48-0081-82) in Flow

          Staining of BALB/c splenocytes with Anti-Mouse CD4 eFluor® 605 and 0.125 µg of Rat IgG2a K Isotype Control eFluor® 450 (Product # 48-4321-82) (left) or 0.125 µg of Anti-Mouse CD8a eFluor® 450 (right). Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Immunocytochemistry (ICC/IF)
          CD8a Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          CD8a Antibody in Immunocytochemistry (ICC/IF)
          CD8a Antibody in Immunocytochemistry (ICC/IF)
          CD8a Antibody in Immunocytochemistry (ICC/IF)
          CD8a Antibody in Immunocytochemistry (ICC/IF)
          CD8a Antibody in Immunocytochemistry (ICC/IF)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry, Flow Cytometry (IHC, Flow)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Immunohistochemistry (IHC)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)
          CD8a Antibody in Flow Cytometry (Flow)

          Product Details

          48-0081-82

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          -
          View 20 publications 20 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 6 publications 6 publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 260 publications 260 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Functional Assay (Functional)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Hamster, Human, Mouse

          Host/Isotype

          Rat / IgG2a, kappa

          Recommended Isotype Control

          Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 450, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          53-6.7

          Conjugate

          eFluor™ 450 eFluor™ 450 eFluor™ 450
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 660 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant UV 661
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 421
          • Brilliant Violet 480
          • Brilliant Violet 605
          • Brilliant Violet 650
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 506
          • eFluor 615
          • eFluor 660
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 585
          • NovaFluor Blue 610-70S
          • NovaFluor Blue 660-120S
          • NovaFluor Blue 725
          • NovaFluor Blue 745
          • NovaFluor Blue 760
          • NovaFluor Blue 800
          • NovaFluor Red 710
          • NovaFluor Red 800
          • NovaFluor UV 765
          • NovaFluor Violet 690
          • NovaFluor Violet 745
          • NovaFluor Violet 800
          • NovaFluor Yellow 590
          • NovaFluor Yellow 610
          • NovaFluor Yellow 660
          • NovaFluor Yellow 690
          • NovaFluor Yellow 700
          • NovaFluor Yellow 730
          • NovaFluor Yellow 810
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          405/445 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1272198

          Product Specific Information

          Description: The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34 kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.

          Applications Reported: This 53-6.7 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 53-6.7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          CD8, also known as cluster of differentiation 8, is a type I transmembrane glycoprotein of the immunoglobulin family that plays a crucial role in T cell differentiation, activation, and signal transduction. It is expressed as either a heterodimer (CD8 alpha beta) or a homodimer (CD8 alpha alpha). The CD8 alpha beta form is predominantly found on the majority of thymocytes and a subpopulation of mature alpha beta TCR T cells, while the CD8 alpha alpha form is expressed on gamma delta TCR T cells, a subset of intestinal intraepithelial lymphocytes (IELs), and dendritic cells. CD8 functions as a co-receptor for major histocompatibility complex class I (MHC-I) molecules, working alongside the T cell receptor (TCR). The CD8 alpha chain is essential for binding to MHC-I. CD8 is also expressed on a subset of T cells, NK cells, monocytes, and dendritic cells as disulfide-linked homodimers of CD8 alpha. Upon ligation of MHC-I/peptide complexes presented by antigen-presenting cells (APCs), CD8 recruits lymphocyte-specific protein tyrosine kinase (Lck), leading to lymphokine production, increased motility, and activation of cytotoxic T lymphocytes (CTLs). Activated CTLs are vital for clearing pathogens and tumor cells. The differentiation of naive CD8+ T cells into CTLs is strongly enhanced by cytokines such as IL-2, IL-12, and TGF-beta1. Through its interactions with MHC-I and association with protein tyrosine kinase p56lck, CD8 plays a significant role in T cell development and the activation of mature T cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: CD8a; CD8alpha; CD8b; CD8beta; fCD8; Leu-2; leu-2a; Lymphocyte antigen 3; Lyt-2.1 lymphocyte differentiation antigen (AA at 100); T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-2; T-cell surface glycoprotein Lyt-3

          View more View less

          Gene Aliases: BB154331; Cd8a; Cd8b; Cd8b1; Ly-2; Ly-3; Ly-35; Ly-B; Ly-C; Lyt-2; Lyt-3; Lyt2; Lyt3

          View more View less

          UniProt ID: (Mouse) P01731, (Mouse) P10300

          View more View less

          Entrez Gene ID: (Mouse) 12525, (Mouse) 12526

          View more View less

          Function(s)
          protein binding protein kinase binding protein homodimerization activity defense/immunity protein immunoglobulin receptor superfamily
          Process(es)
          immune system process T cell mediated immunity cell surface receptor signaling pathway T cell activation cytotoxic T cell differentiation positive regulation of calcium-mediated signaling defense response to virus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline