Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • FcRL5 Antibodies

          Invitrogen

          CD307e (FcRL5) Monoclonal Antibody (509F6), eFluor™ 660, eBioscience™

          2 References
          View all (11) FcRL5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD307e (FcRL5) Monoclonal Antibody (509F6), eFluor™ 660, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CD307e (FcRL5) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD307e (FcRL5) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD307e (FcRL5) Antibody (50-3078-42) in Flow

          Staining of normal human peripheral blood cells (left) and human tonsil cells (right) with Anti-Human CD19 eFluor® 450 (Product # 48-0198-42) and Anti-Human CD307e (FcRL5) eFluor® 660 (Product # 50-3078-42). Cells in the lymphocyte gate were used for analysis. Data courtesy of the laboratory of Shane Crotty. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD307e (FcRL5) Antibody in Flow Cytometry (Flow)

          Product Details

          50-3078-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 µL (1 µg)/test
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG2a, kappa

          Recommended Isotype Control

          Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 660, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          509F6

          Conjugate

          eFluor™ 660 eFluor™ 660 eFluor™ 660

          Excitation/Emission Max

          651/668 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 µL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2574199

          Product Specific Information

          Description: The 509F6 monoclonal antibody reacts with human CD307e which is also known as Fc receptor-like 5, FcRL5, FcRH5, or IRTA2. CD307e is a surface protein expressed selectively on naive mature B cells and on bone marrow plasma cells, and binds to all IgG isotypes. CD307e functions as an inhibitory coreceptor on mature B cells in humans via the recruitment of the phosphatase SHP-1 to its two immunoreceptor tyrosine-based inhibitory motifs (ITIM). CD307e can also exist in soluble form and high levels of soluble CD307e have been detected in the blood of patients with multiple myeloma, chronic lymphocytic leukemia, and mantle cell lymphoma. Because surface-bound CD307e is also highly-expressed on malignant B cells from multiple myeloma, chronic lymphocytic leukemia, and mantle cell lymphoma patients, CD307e may be a useful therapeutic target for the treatment of these cancers.

          Applications Reported: This 509F6 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 509F6 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (1 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Fc receptor homologs (FcRH5), also known as IRTA2 proteins, are transmembrane proteins with sequence homology to classical Fc receptor families. FcRH5 function as activating or inhibitory receptors on discrete B cell populations and are involved in B-cell development and differentiation in peripheral lymphoid organs. FcRH5 expression is deregulated in multiple myeloma and Burkitt lymphoma cell lines with 1q21 abnormalities. FcRH5 may play a critical role for the pathogenesis of B-cell lymphoma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: BXMAS1; CD307e; Fc receptor homolog 5; Fc receptor-like protein 5; FcR-like protein 5; FcRH5; Ig-like superfamily receptor; Immune receptor translocation-associated protein 2; immunoglobulin superfamily receptor translocation associated 2 (IRTA2); immunoglobulin-like domain and ITIM motif-containing receptor; IRTA2a; IRTA2b; IRTA2c; IRTA2d; unnamed protein product

          View more View less

          Gene Aliases: BXMAS1; CD307; CD307e; FCRH5; FCRL5; IRTA2; PRO820; UNQ503/PRO820

          View more View less

          UniProt ID: (Human) Q96RD9

          View more View less

          Entrez Gene ID: (Human) 83416

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding coreceptor activity
          Process(es)
          immune response cell surface receptor signaling pathway single fertilization B cell activation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline