Product References
Controlled X-chromosome dynamics defines meiotic potential of female mouse in vitro germ cells.
The EMBO journal
Severino J,Bauer M,Mattimoe T,Arecco N,Cozzuto L,Lorden P,Hamada N,Nosaka Y,Nagaoka SI,Audergon P,Tarruell A,Heyn H,Hayashi K,Saitou M,Payer B
50-8813-42 was used in Flow cytometry/Cell sorting to investigate X-inactivation and reactivation dynamics using a tailor-made in vitro system of primordial germ cell-like cell (PGCLC) differentiation from mouse embryonic stem cells.
Tue Jun 14 00:00:00 EDT 2022
Disruption of the MSL complex inhibits tumour maintenance by exacerbating chromosomal instability.
Nature cell biology
Monserrat J,Morales Torres C,Richardson L,Wilson TS,Patel H,Domart MC,Horswell S,Song OR,Jiang M,Crawford M,Bui M,Dalal Y,Scaffidi P
50-8813-42 was used in Flow Cytometry to demonstrate that targeting MSL (male-specific lethal complex) may be a valuable means to increase CIN (chromosomal instability) beyond the level tolerated by cancer cells without inducing severe adverse effects in normal tissues.
Thu Apr 01 00:00:00 EDT 2021
High-throughput screening in postimplantation haploid epiblast stem cells reveals Hs3st3b1 as a modulator for reprogramming.
Stem cells translational medicine
Gao Q,Zhang W,Zhao Y,Tian Y,Wang Y,Zhang J,Geng M,Xu M,Yao C,Wang H,Li L,Liu Y,Shuai L
50-8813-42 was used in Flow Cytometry to open the feasibility of utilizing haEpiSCs to elucidate fundamental biological processes including cell fate alternations.
Sat May 01 00:00:00 EDT 2021
High-throughput screening in postimplantation haploid epiblast stem cells reveals Hs3st3b1 as a modulator for reprogramming.
Stem cells translational medicine
Gao Q,Zhang W,Zhao Y,Tian Y,Wang Y,Zhang J,Geng M,Xu M,Yao C,Wang H,Li L,Liu Y,Shuai L
50-8813-42 was used in Flow Cytometry to open the feasibility of utilizing haEpiSCs to elucidate fundamental biological processes including cell fate alternations.
Sat May 01 00:00:00 EDT 2021
Protocols for analysis of mitochondrial permeability transition pore opening in mouse somatic cell reprogramming.
STAR protocols
Ying Z,Liu Z,Xiang G,Xin Y,Wang J,Liu X
50-8813-42 was used in Flow Cytometry to describe a protocol for imaging mitochondrial permeability transition pore (mPTP) opening in reprogramming of somatic cells using a confocal microscope.
Fri Jun 18 00:00:00 EDT 2021
Chromosome compartments on the inactive X guide TAD formation independently of transcription during X-reactivation.
Nature communications
Bauer M,Vidal E,Zorita E,Üresin N,Pinter SF,Filion GJ,Payer B
50-8813-42 was used in Flow Cytometry to investigate if topologically associating domains formation and transcriptional reactivation are causally independent during X-reactivation while establishing Xist as a common denominator.
Wed Jun 09 00:00:00 EDT 2021
ECE2 regulates neurogenesis and neuronal migration during human cortical development.
EMBO reports
Buchsbaum IY,Kielkowski P,Giorgio G,O'Neill AC,Di Giaimo R,Kyrousi C,Khattak S,Sieber SA,Robertson SP,Cappello S
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Wed May 06 00:00:00 EDT 2020
Functional Oocytes Derived from Granulosa Cells.
Cell reports
Tian C,Liu L,Ye X,Fu H,Sheng X,Wang L,Wang H,Heng D,Liu L
50-8813-42 was used in Flow cytometry/Cell sorting to demonstrate that funtional oocytes posessing high genomic stability can be produced in vitro from somatic gPSCs through cellular reprogramming, facilitated by protonic sodium or acid - overall a safe and effective chemical induction technique.
Tue Dec 24 00:00:00 EST 2019
Comparative glycoproteomics of stem cells identifies new players in ricin toxicity.
Nature
Stadlmann J,Taubenschmid J,Wenzel D,Gattinger A,Dürnberger G,Dusberger F,Elling U,Mach L,Mechtler K,Penninger JM
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunocytochemistry
Thu Sep 28 00:00:00 EDT 2017
DND1 maintains germline stem cells via recruitment of the CCR4-NOT complex to target mRNAs.
Nature
Yamaji M,Jishage M,Meyer C,Suryawanshi H,Der E,Yamaji M,Garzia A,Morozov P,Manickavel S,McFarland HL,Roeder RG,Hafner M,Tuschl T
50-8813 was used in Flow cytometry/Cell sorting to investigate the molecular function of DND1 protein.
Thu Mar 23 00:00:00 EDT 2017
A vital sugar code for ricin toxicity.
Cell research
Taubenschmid J,Stadlmann J,Jost M,Klokk TI,Rillahan CD,Leibbrandt A,Mechtler K,Paulson JC,Jude J,Zuber J,Sandvig K,Elling U,Marquardt T,Thiel C,Koerner C,Penninger JM
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Wed Nov 01 00:00:00 EDT 2017
A vital sugar code for ricin toxicity.
Cell research
Taubenschmid J,Stadlmann J,Jost M,Klokk TI,Rillahan CD,Leibbrandt A,Mechtler K,Paulson JC,Jude J,Zuber J,Sandvig K,Elling U,Marquardt T,Thiel C,Koerner C,Penninger JM
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Wed Nov 01 00:00:00 EDT 2017
Not All H3K4 Methylations Are Created Equal: Mll2/COMPASS Dependency in Primordial Germ Cell Specification.
Molecular cell
Hu D,Gao X,Cao K,Morgan MA,Mas G,Smith ER,Volk AG,Bartom ET,Crispino JD,Di Croce L,Shilatifard A
50-8813 was used in Flow cytometry/Cell sorting to analyse the contribution of MLL2's methyltransferase and CXXC domain in H3K4 trimethylation in mouse embryonic stem cells.
Thu Feb 02 00:00:00 EST 2017
Not All H3K4 Methylations Are Created Equal: Mll2/COMPASS Dependency in Primordial Germ Cell Specification.
Molecular cell
Hu D,Gao X,Cao K,Morgan MA,Mas G,Smith ER,Volk AG,Bartom ET,Crispino JD,Di Croce L,Shilatifard A
50-8813 was used in Flow cytometry/Cell sorting to analyse the contribution of MLL2's methyltransferase and CXXC domain in H3K4 trimethylation in mouse embryonic stem cells.
Thu Feb 02 00:00:00 EST 2017
A molecular roadmap of the AGM region reveals BMPER as a novel regulator of HSC maturation.
The Journal of experimental medicine
McGarvey AC,Rybtsov S,Souilhol C,Tamagno S,Rice R,Hills D,Godwin D,Rice D,Tomlinson SR,Medvinsky A
50-8813-42 was used in Immunohistochemistry to identify novel secreted regulators of haematopoietic stem cell maturation.
Mon Dec 04 00:00:00 EST 2017
Noncanonical function of DGCR8 controls mESC exit from pluripotency.
The Journal of cell biology
Cirera-Salinas D,Yu J,Bodak M,Ngondo RP,Herbert KM,Ciaudo C
50-8813 was used in Flow cytometry/Cell sorting to reveal a new noncanonical function of DGCR8 in the modulation of the alternative splicing of Tcf7l1 mRNA in addition to its established function in microRNA biogenesis.
Wed Feb 01 00:00:00 EST 2017
NANOG alone induces germ cells in primed epiblast in vitro by activation of enhancers.
Nature
Murakami K,Günesdogan U,Zylicz JJ,Tang WWC,Sengupta R,Kobayashi T,Kim S,Butler R,Dietmann S,Surani MA
50-8813 was used in Flow cytometry/Cell sorting to investigate the role of nanog in unipotent primordial germ cells in mice.
Thu Jan 21 00:00:00 EST 2016
NANOG alone induces germ cells in primed epiblast in vitro by activation of enhancers.
Nature
Murakami K,Günesdogan U,Zylicz JJ,Tang WWC,Sengupta R,Kobayashi T,Kim S,Butler R,Dietmann S,Surani MA
50-8813 was used in Flow cytometry/Cell sorting to investigate the role of nanog in unipotent primordial germ cells in mice.
Thu Jan 21 00:00:00 EST 2016
Foxd3 Promotes Exit from Naive Pluripotency through Enhancer Decommissioning and Inhibits Germline Specification.
Cell stem cell
Respuela P,Nikolić M,Tan M,Frommolt P,Zhao Y,Wysocka J,Rada-Iglesias A
50-8813 was used in Flow cytometry/Cell sorting to discover a cycle of activation and deactivation of Foxd3 required for exit from naive pluripotency and primordial germ cell specification.
Thu Jan 07 00:00:00 EST 2016
The transcriptomes of novel marmoset monkey embryonic stem cell lines reflect distinct genomic features.
Scientific reports
Debowski K,Drummer C,Lentes J,Cors M,Dressel R,Lingner T,Salinas-Riester G,Fuchs S,Sasaki E,Behr R
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunohistochemistry
Thu Jul 07 00:00:00 EDT 2016
Alternative Routes to Induced Pluripotent Stem Cells Revealed by Reprogramming of the Neural Lineage.
Stem cell reports
Jackson SA,Olufs ZP,Tran KA,Zaidan NZ,Sridharan R
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Tue Mar 08 00:00:00 EST 2016
Parent stem cells can serve as niches for their daughter cells.
Nature
Pardo-Saganta A,Tata PR,Law BM,Saez B,Chow RD,Prabhu M,Gridley T,Rajagopal J
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Thu Jul 30 00:00:00 EDT 2015
Parent stem cells can serve as niches for their daughter cells.
Nature
Pardo-Saganta A,Tata PR,Law BM,Saez B,Chow RD,Prabhu M,Gridley T,Rajagopal J
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Thu Jul 30 00:00:00 EDT 2015
Parent stem cells can serve as niches for their daughter cells.
Nature
Pardo-Saganta A,Tata PR,Law BM,Saez B,Chow RD,Prabhu M,Gridley T,Rajagopal J
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Thu Jul 30 00:00:00 EDT 2015
An alternative pluripotent state confers interspecies chimaeric competency.
Nature
Wu J,Okamura D,Li M,Suzuki K,Luo C,Ma L,He Y,Li Z,Benner C,Tamura I,Krause MN,Nery JR,Du T,Zhang Z,Hishida T,Takahashi Y,Aizawa E,Kim NY,Lajara J,Guillen P,Campistol JM,Esteban CR,Ross PJ,Saghatelian A,Ren B,Ecker JR,Izpisua Belmonte JC
50-8813 was used in Flow cytometry/Cell sorting to show modulation of culture parameters efficiently selects for region-selective pluripotent stem cells from mouse embryos and primate pluripotent stem cells.
Thu May 21 00:00:00 EDT 2015
Injury induces direct lineage segregation of functionally distinct airway basal stem/progenitor cell subpopulations.
Cell stem cell
Pardo-Saganta A,Law BM,Tata PR,Villoria J,Saez B,Mou H,Zhao R,Rajagopal J
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Immunofluorescence
Thu Feb 05 00:00:00 EST 2015
Coexpression analysis of CD133 and CD44 identifies proneural and mesenchymal subtypes of glioblastoma multiforme.
Oncotarget
Brown DV,Daniel PM,D'Abaco GM,Gogos A,Ng W,Morokoff AP,Mantamadiotis T
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Fri Mar 20 00:00:00 EDT 2015
Distinct and combinatorial functions of Jmjd2b/Kdm4b and Jmjd2c/Kdm4c in mouse embryonic stem cell identity.
Molecular cell
Das PP,Shao Z,Beyaz S,Apostolou E,Pinello L,De Los Angeles A,O'Brien K,Atsma JM,Fujiwara Y,Nguyen M,Ljuboja D,Guo G,Woo A,Yuan GC,Onder T,Daley G,Hochedlinger K,Kim J,Orkin SH
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Thu Jan 09 00:00:00 EST 2014
Dax1 and Nanog act in parallel to stabilize mouse embryonic stem cells and induced pluripotency.
Nature communications
Zhang J,Liu G,Ruan Y,Wang J,Zhao K,Wan Y,Liu B,Zheng H,Peng T,Wu W,He P,Hu FQ,Jian R
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Mon Oct 06 00:00:00 EDT 2014
Cell reprogramming requires silencing of a core subset of polycomb targets.
PLoS genetics
Fragola G,Germain PL,Laise P,Cuomo A,Blasimme A,Gross F,Signaroldi E,Bucci G,Sommer C,Pruneri G,Mazzarol G,Bonaldi T,Mostoslavsky G,Casola S,Testa G
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Thu Jun 13 00:00:00 EDT 2013
Cell reprogramming requires silencing of a core subset of polycomb targets.
PLoS genetics
Fragola G,Germain PL,Laise P,Cuomo A,Blasimme A,Gross F,Signaroldi E,Bucci G,Sommer C,Pruneri G,Mazzarol G,Bonaldi T,Mostoslavsky G,Casola S,Testa G
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Thu Jun 13 00:00:00 EDT 2013
Cell reprogramming requires silencing of a core subset of polycomb targets.
PLoS genetics
Fragola G,Germain PL,Laise P,Cuomo A,Blasimme A,Gross F,Signaroldi E,Bucci G,Sommer C,Pruneri G,Mazzarol G,Bonaldi T,Mostoslavsky G,Casola S,Testa G
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Thu Jun 13 00:00:00 EDT 2013
Embryonic stem cells require Wnt proteins to prevent differentiation to epiblast stem cells.
Nature cell biology
ten Berge D,Kurek D,Blauwkamp T,Koole W,Maas A,Eroglu E,Siu RK,Nusse R
Published figure using SSEA1 monoclonal antibody (Product # 50-8813-42) in Flow Cytometry
Sun Aug 14 00:00:00 EDT 2011