Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CCL5 (RANTES) Antibodies

          Invitrogen

          CCL5 (RANTES) Polyclonal Antibody, PeproTech®

          2 Published Figures
          11 References
          View all (30) CCL5 (RANTES) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CCL5 (RANTES) Polyclonal Antibody, PeproTech®

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (2)
          CCL5 (RANTES) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CCL5 (RANTES) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CCL5 (RANTES) Antibody (500-P36-1MG) in WB

          Western Blot: To detect hRANTES by Western Blot analysis CCL5 (RANTES) Polyclonal Antibody (Product # 500-P36-1MG) can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hRANTES is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CCL5 (RANTES) Antibody in Western Blot (WB)
          CCL5 (RANTES) Antibody in Western Blot (WB)
          CCL5 (RANTES) Antibody in ELISA (ELISA)
          CCL5 (RANTES) Antibody in Neutralization (Neu)
          CCL5 (RANTES) Antibody in Neutralization (Neu)
          CCL5 (RANTES) Polyclonal Antibody, PeproTech®

          Product Details

          500-P36-1MG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.2 µg/mL
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.5-2.0 µg/mL
          View 2 publications 2 publications

          Neutralization (Neu)

          -
          View 2 publications 2 publications

          In vitro Assay (In vitro)

          -
          View 1 publication 1 publication

          Immunostaining (IS)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human

          Published species

          Bacteria, Human

          Host/Isotype

          Rabbit

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived Recombinant Human RANTES (CCL5)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.1-1.0 mg/mL

          Amount

          1 mg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2930486

          Product Specific Information

          AA Sequence of recombinant protein: SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS.

          Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human RANTES (CCL5). Anti-Human RANTES (CCL5)-specific antibody was purified by affinity chromatography employing an immobilized Human RANTES (CCL5) matrix.

          Sandwich ELISA: To detect Human RANTES (CCL5) by sandwich ELISA (using 100 µL/well antibody solution) a concentration of 0.5-2.0 µg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human RANTES (CCL5) (500-P36Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human RANTES (CCL5).

          Western Blot: To detect hRANTES by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 µg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hRANTES is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

          Target Information

          CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: beta-chemokine RANTES; C-C motif chemokine 5; chemokine (C-C motif) ligand 5; EoCP; Eosinophil chemotactic cytokine; MGC17164; OTTHUMP00000197106; regulated upon activation, normally T-expressed, and presumably secreted; SIS δ; SIS-delta; SISδ; small inducible cytokine subfamily A (Cys-Cys), member 5; Small-inducible cytokine A5; T cell-specific protein P228; T cell-specific protein precursor; T-cell specific protein p288; T-cell-specific protein RANTES; TCP228; unnamed protein product

          View more View less

          Gene Aliases: CCL5; D17S136E; eoCP; RANTES; SCYA5; SIS-delta; SISd; TCP228

          View more View less

          UniProt ID: (Human) P13501

          View more View less

          Entrez Gene ID: (Human) 6352

          View more View less

          Function(s)
          phosphatidylinositol phospholipase C activity protein kinase activity cytokine activity protein binding chemokine activity phospholipase activator activity receptor signaling protein tyrosine kinase activator activity CCR1 chemokine receptor binding CCR4 chemokine receptor binding CCR5 chemokine receptor binding chemoattractant activity chemokine receptor binding identical protein binding protein homodimerization activity chemokine receptor antagonist activity CCR chemokine receptor binding
          Process(es)
          dendritic cell chemotaxis monocyte chemotaxis regulation of chronic inflammatory response calcium ion transport cellular calcium ion homeostasis exocytosis chemotaxis inflammatory response immune response leukocyte cell-cell adhesion G-protein coupled receptor signaling pathway cell-cell signaling response to virus response to toxic substance positive regulation of macrophage chemotaxis positive regulation of T cell chemotaxis positive regulation of smooth muscle cell migration positive regulation of cell migration phospholipase D-activating G-protein coupled receptor signaling pathway positive regulation of TOR signaling positive regulation of cell-cell adhesion mediated by integrin response to cytokine positive regulation of homotypic cell-cell adhesion response to tumor necrosis factor chemokine (C-C motif) ligand 5 signaling pathway positive regulation of T cell proliferation neutrophil activation negative regulation by host of viral transcription cellular response to fibroblast growth factor stimulus positive regulation of viral genome replication negative regulation of viral genome replication positive regulation of innate immune response negative regulation of G-protein coupled receptor protein signaling pathway positive regulation of G-protein coupled receptor protein signaling pathway positive regulation of cell adhesion positive regulation of translational initiation eosinophil chemotaxis macrophage chemotaxis positive regulation of smooth muscle cell proliferation epithelial cell proliferation positive regulation of epithelial cell proliferation regulation of insulin secretion regulation of T cell activation positive chemotaxis positive regulation of protein kinase B signaling positive regulation of calcium ion transport antimicrobial humoral immune response mediated by antimicrobial peptide chemokine-mediated signaling pathway negative regulation of chemokine-mediated signaling pathway negative regulation of T cell apoptotic process positive regulation of T cell apoptotic process ERK1 and ERK2 cascade cellular response to interferon-gamma cellular response to interleukin-1 cellular response to tumor necrosis factor positive regulation of monocyte chemotaxis receptor signaling pathway via STAT cellular response to virus positive regulation of neuroinflammatory response positive regulation of STAT cascade negative regulation of macrophage apoptotic process positive regulation of T cell migration positive regulation of natural killer cell chemotaxis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline