Search
Search
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene is a member of the paired box (PAX) family of transcription factors. Members of this gene family typically contain a paired box domain, an octapeptide, and a paired-type homeodomain. These genes play critical roles during fetal development and cancer growth. The specific function of the paired box 7 gene is unknown but speculated to involve tumor suppression since fusion of this gene with a forkhead domain family member has been associated with alveolar rhabdomyosarcoma. Three transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ37460; HuP1; paired box gene 7; paired box homeotic gene 7; Paired box protein Pax-7; paired box transcription factor PAX7; paired domain gene 7; PAX7 transcriptional factor; PAX7A; PAX7B
Gene Aliases: CMYO19; CMYP19; HUP1; MYOSCO; PAX-7; PAX7; PAX7B; RGD1564360; RMS2
UniProt ID: (Human) P23759, (Mouse) P47239
Entrez Gene ID: (Chicken) 395942, (Human) 5081, (Mouse) 18509, (Rat) 500574
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support