Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • PSD-95 Antibodies

          Invitrogen

          PSD-95 Polyclonal Antibody

          29 Published Figures
          132 References
          View all (56) PSD-95 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PSD-95 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Published Figures (29)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 31

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PSD-95 Antibody (51-6900) in ICC/IF

          Immunofluorescent analysis of PSD-95 (green) and MAP2 (red) on rat primary cortical neurons cultured for 28 days in the B-27 Plus Neuronal Culture System (Product # A3653401). At day 28 the cells were fixed with 4% paraformaldehyde for 15 min, permeabilized with 0.1% triton X-100 for 30min, and blocked with 1% BSA for 30 min at room temperature. Cells were stained with anti-PSD95 antibody (Product # 51-6900) at a dilution of 1:200, and anti-MAP2 (Product # 13-1500) at a dilution of 1:400, in 1% BSA staining buffer, overnight at 4C, and the... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PSD-95 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunocytochemistry (ICC/IF)
          PSD-95 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PSD-95 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry (IHC)
          PSD-95 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          PSD-95 Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Antibody in Western Blot (WB)
          PSD-95 Polyclonal Antibody

          Product Details

          51-6900

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1.0 µg/mL
          View 24 publications 24 publications

          Immunohistochemistry (IHC)

          -
          View 67 publications 67 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 10 publications 10 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 3 publications 3 publications

          Immunocytochemistry (ICC/IF)

          1:200
          View 23 publications 23 publications

          ELISA (ELISA)

          0.1-1.0 µg/mL
          -

          Immunoprecipitation (IP)

          5 µg
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Chicken, Human, Mouse, Rat, Xenopus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH-conjugated synthethic peptide derived from the C-terminal region of human PSD-95.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533914

          Product Specific Information

          This antibody recognizes the 95 kDa PSD-95 protein. Cross-reactivity with an unknown protein at ~50 kDa has been observed in western blots with rat brain homogenates. Western blots with mouse brain homogenates did not show such a band.

          Positive reactivity of this antibody was confirmed on western blots of rat brain homogenates and lysates of PSD-95- transfected HEK293 (human embryonic kidney) cells.

          Target Information

          Post Synaptic Density 95 kDa (PSD-95), also known as synapse associated protein 90 kDa (SAP90), is one of a family of membrane-associated proteins found in the postsynaptic density in forebrain neurons and certain presynaptic structures in the cerebellum. Like other members of the family, PSD-95 has three 90 amino acid repeats called PDZ domains followed by an SH3 domain and a yeast guanylate kinase homology (GuK) domain. PSD-95 is believed to participate in the clustering of certain proteins, including NMDA receptors, Shaker-type potassium channels at the synaptic membrane in central nervous system (CNS) neurons. There are two principal modes of interaction between PSD-95 and other proteins. NMDA receptors and shaker-type potassium channels both share C-terminal sequence homology consisting of a threonine/serine-X-valine-COOH (T/SXV) motif. Other neuronal proteins that share this motif (beta 1 adrenergic receptor, some serotonin receptors, some sodium channel subunits, and additional potassium channel subunits), and some of these proteins may interact with PSD-95 by binding to its PDZ domains. Neuronal nitric oxide synthase (nNOS), which lacks the T/SXV motif but which has its own PDZ domain, has been shown to associate with PSD-95 in vitro through a pseudo-homotypic PDZ-PDZ interaction.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: discs large homolog 4; Disks large homolog 4; FLJ97752; FLJ98574; post synaptic density; post-synaptic density protein 95; Postsynaptic density protein 95; PSD-95; PSD-95 alpha 2b; PSD-95 beta; SAP-90; Synapse-associated protein 90; synapse-associated protein SAP90; Tax interaction protein 15; unnamed protein product

          View more View less

          Gene Aliases: DLG4; Dlgh4; MRD62; PSD-95; PSD95; SAP-90; SAP90; SAP90A

          View more View less

          UniProt ID: (Human) P78352, (Rat) P31016, (Mouse) Q62108

          View more View less

          Entrez Gene ID: (Human) 1742, (Rat) 29495, (Mouse) 13385

          View more View less

          Function(s)
          protein binding kinase binding protein kinase binding protein phosphatase binding PDZ domain binding protein binding, bridging beta-1 adrenergic receptor binding D1 dopamine receptor binding P2Y1 nucleotide receptor binding acetylcholine receptor binding ionotropic glutamate receptor binding macromolecular complex binding neuroligin family protein binding scaffold protein binding guanylate kinase activity receptor binding protein C-terminus binding protein complex binding glutamate receptor binding structural molecule activity neurexin family protein binding scaffold/adaptor protein
          Process(es)
          negative regulation of receptor internalization signal transduction positive regulation of cytosolic calcium ion concentration chemical synaptic transmission nervous system development learning synaptic vesicle maturation social behavior protein localization to synapse locomotory exploration behavior cellular response to potassium ion establishment of protein localization establishment or maintenance of epithelial cell apical/basal polarity regulation of long-term neuronal synaptic plasticity positive regulation of synaptic transmission neuromuscular process controlling balance dendritic spine morphogenesis protein-containing complex assembly vocalization behavior AMPA glutamate receptor clustering receptor localization to synapse cell-cell adhesion NMDA selective glutamate receptor signaling pathway regulation of postsynaptic membrane neurotransmitter receptor levels neurotransmitter receptor localization to postsynaptic specialization membrane positive regulation of neuron projection arborization positive regulation of excitatory postsynaptic potential regulation of grooming behavior protein complex assembly neuronal ion channel clustering nucleotide phosphorylation regulation of N-methyl-D-aspartate selective glutamate receptor activity locomotory behavior response to cocaine regulation of neuronal synaptic plasticity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline