Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: MTKKSAAPT DRQPYSLCSN RKSLSQQLDC PAGKAAGTSR PTRSLSTAQL VQPSGGLQAS VISNIVLMKG QAKGLGFSIV GGKDSIYGPI GIYVKTIFAG GAAAADGRLQ EGDEILELNG ESMAGLTHQD ALQKFKQAKK GLLTLTVRTR LTAPPSLCSH LSPPLCRSLS SSTCITKDSS SFALESPSAP ISTAKPNYRI MVEVSLQKEA GVGLGIGLCS VPYFQCISGI FVHTLSPGSV AHLDGRLRCG DEIVEISDSP VHCLTLNEVY TILSHCDPGP VSIIVSRHPD PQKEEMEAQE VK (154-454 aa encoded by BC040272)
The protein encoded by IL-16 is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of IL-16 undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Alternate splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; ILN; Interleukin; Interleukin16; lymphocyte chemoattractant factor; M-IL-16; neuronal interleukin 16; Pro-interleukin-16; prointerleukin 16
Gene Aliases: IL16; LCF; NIL16; prIL-16; PRIL16
UniProt ID: (Human) Q14005
Entrez Gene ID: (Human) 3603
Molecular Function: interleukin superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support