Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • alpha Catenin Antibodies

          Invitrogen

          alpha Catenin Polyclonal Antibody

          Advanced Verification
          9 Published Figures
          28 References
          View all (11) alpha Catenin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite alpha Catenin Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (9)
          • Advanced Verification (2)
          alpha Catenin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          alpha Catenin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          alpha Catenin Antibody (71-1200) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of A431 (Lane 1), A549 (Lane 2), CaCo2 (Lane 3), MCF7 (lane 4) and HT-29 (lane 5). The blots were probed with Anti-alpha Catenin Rabbit Polyclonal Antibody (Product # 71-1200, 1-2 µg/mL) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A27036, 0.4 µg/mL, 1:2500 dilution). Two bands ~ 100 and 59 kDa corresponding to alpha Catenin was observed across cell lines tested. Known quantity of... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Western Blot (WB)
          alpha Catenin Antibody in Immunoprecipitation (IP)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          alpha Catenin Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunocytochemistry (ICC/IF)
          alpha Catenin Antibody in Immunohistochemistry (IHC)
          alpha Catenin Antibody
          alpha Catenin Antibody

          Product Details

          71-1200

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          View 11 publications 11 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 10 publications 10 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse, Rat, Xenopus

          Published species

          Bovine, Dog, Hamster, Human, Mouse, Rat, Virus

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Human alpha Catenin

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533974

          Target Information

          Alpha catenin is a member of the b-catenin family of cytosolic proteins. Alpha catenin interacts with N-cadherin and b-catenin on the cytosolic side of adherins junctions to mediate the interaction with the actin cytoskeleton. Alpha catenin is a homodimer in solution, but dissociates to form a 1:1 heterodimer with b-catenin in order to interact with cadherins. It also regulates morphological plasticity of synapses and cerebellar and hippocampal lamination during development. Alpha catenin also functions in the control of the startle modulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 102 kDa cadherin-associated protein; alpha E catenin; Alpha E-catenin; Alpha N-catenin; alpha(E)-catenin; alpha(N)-catenin; alpha-catenin; Alpha-catenin-related protein; alpha-E-catenin; alpha-N-catenin; alphaE-catenin; alphaN-catenin; cadherin associated protein; Cadherin-associated protein; cadherin-associated protein, related; cancer/testis antigen 114; CAP102; catenin; catenin (cadherin associated protein), alpha 1; catenin (cadherin-associated protein), alpha 1, 102kDa; catenin (cadherin-associated protein), alpha 2; catenin alpha 1; catenin alpha 2; Catenin alpha-1; Catenin alpha-2; Catenin alpha1; Catenin alpha2; catenin, alpha 1; Catenin-alpha1; Catenin-alpha2; cerebellar deficient folia; epididymis secretory sperm binding protein; FLJ36832; FLJ52416; HGNC:2509; Neural alpha-catenin; NY-REN-13 antigen; Renal carcinoma antigen NY-REN-13; unnamed protein product

          View more View less

          Gene Aliases: 2010010M04Rik; AA517462; AI481747; AI988031; CAP-R; CAP102; CAPR; Catna; Catna1; Catna2; CDCBM9; cdf; chp; CT114; CTNNA1; CTNNA2; CTNR; MDBS2; MDPT2

          View more View less

          UniProt ID: (Human) P35221, (Chicken) P30997, (Human) P26232, (Mouse) P26231, (Mouse) Q61301

          View more View less

          Entrez Gene ID: (Chicken) 416184, (Human) 1495, (Chicken) 396035, (Human) 1496, (Rat) 307505, (Mouse) 12385, (Mouse) 12386, (Rat) 297357

          View more View less

          Function(s)
          RNA binding structural molecule activity protein binding beta-catenin binding cytoskeletal protein binding vinculin binding identical protein binding macromolecular complex binding gamma-catenin binding cadherin binding actin filament binding structural constituent of cytoskeleton poly(A) RNA binding non-motor actin binding protein
          Process(es)
          ovarian follicle development apoptotic process cell adhesion establishment or maintenance of cell polarity smoothened signaling pathway integrin-mediated signaling pathway neuroblast proliferation negative regulation of neuroblast proliferation protein localization male gonad development gap junction assembly cell migration axon regeneration regulation of cell proliferation odontogenesis of dentin-containing tooth negative regulation of apoptotic process apical junction assembly response to estrogen positive regulation of smoothened signaling pathway cell motility cellular response to indole-3-methanol epithelial cell-cell adhesion extrinsic apoptotic signaling pathway in absence of ligand cell-cell adhesion negative regulation of protein localization to nucleus negative regulation of cell motility negative regulation of integrin-mediated signaling pathway negative regulation of extrinsic apoptotic signaling pathway in absence of ligand positive regulation of extrinsic apoptotic signaling pathway in absence of ligand neural crest cell migration axonogenesis epithelial cell differentiation dendrite morphogenesis brain morphogenesis regulation of synapse structural plasticity trigeminal ganglion formation cytoskeleton organization regulation of neuron projection development radial glia guided migration of Purkinje cell cell differentiation negative regulation of Arp2/3 complex-mediated actin nucleation prepulse inhibition modification of postsynaptic actin cytoskeleton regulation of neuron migration actin filament organization aging response to organic cyclic compound cellular protein localization protein heterooligomerization multicellular organism development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline