Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PAX2 Antibodies

          Invitrogen

          PAX2 Polyclonal Antibody

          70 Published Figures
          183 References
          View all (39) PAX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite PAX2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (70)
          PAX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PAX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 71

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PAX2 Antibody (71-6000) in WB

          Western blot of mouse embryonic kidney using Rabbit anti-Pax-2 Polyclonal Antibody (Product # 71-6000). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          PAX2 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunocytochemistry (ICC/IF)
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Immunohistochemistry (IHC)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in Western Blot (WB)
          PAX2 Antibody in In Situ Hybridization (ISH)
          PAX2 Polyclonal Antibody

          Product Details

          71-6000

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 13 publications 13 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 97 publications 97 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 21 publications 21 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          - -

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 11 publications 11 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 28 publications 28 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 10 publications 10 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Sheep, Rat, Xenopus, Zebrafish

          Published species

          Chicken, Cynomolgus monkey, Dog, Human, Mouse, Rat, Rhesus monkey, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A GST-Pax-2 fusion protein derived from the C-terminal domain (aa188-385) of the murine Pax-2 protein
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533990

          Target Information

          The pax genes are a family of transcription factors that are active in specific tissues during early embryonic development. Pax family members possess a DNA-binding domain encoded by the paired box. Because paired domain containing genes encode transcription factors, they are capable of executing a genetic program. Several pax genes have also been associated with developmental mutations including: pax-3, which is associated with Waardenburg syndrome, pax-6, which is associated with Aniridia, and pax-2, which is associated with Wilms tumor. During embryogenesis, Pax-2 is expressed in the developing kidney. In particular, the pax-2 gene is expressed in condensing metanephric mesenchyme and in early epithelial structures derived from mesenchyme; however, pax-2 mRNA and protein levels are rapidly down regulated as the tubular epithelium matures. Although Pax-2 is down regulated during renal epithelium maturation, Pax-2 expression persists in the undifferentiated epithelium of Wilms' tumors. Persistent expression of Pax-2 in Wilms' tumors occurs frequently and correlates with the proliferation of poorly differentiated epithelial cells in these tumors. Interestingly, expression of the Wilms' tumor suppresser protein, WT1, coincides with down-regulation of the pax-2 gene; thus, suggesting that WT1 can directly repress pax-2 transcription.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: optic disc coloboma; paired box gene 2; paired box homeotic gene 2; Paired box protein Pax-2

          View more View less

          Gene Aliases: FSGS7; Opdc; PAPRS; PAX-2; PAX2

          View more View less

          UniProt ID: (Human) Q02962, (Mouse) P32114

          View more View less

          Entrez Gene ID: (Sheep) 100422802, (Human) 5076, (Mouse) 18504, (Rat) 293992

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding core promoter proximal region sequence-specific DNA binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding transcription factor binding sequence-specific double-stranded DNA binding transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding superoxide-generating NADPH oxidase activity transcription regulatory region DNA binding C2H2 zinc finger domain binding sequence-specific DNA binding
          Process(es)
          urogenital system development branching involved in ureteric bud morphogenesis cell fate determination mesonephros development neural tube closure optic cup morphogenesis involved in camera-type eye development mesenchymal to epithelial transition involved in metanephros morphogenesis retinal pigment epithelium development regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter nervous system development axonogenesis sensory organ development mesodermal cell fate specification visual perception glial cell differentiation optic nerve development optic nerve morphogenesis optic nerve structural organization vestibulocochlear nerve formation cell differentiation regulation of metanephros size ureter maturation pronephric field specification inner ear morphogenesis camera-type eye development negative regulation of apoptotic process negative regulation of programmed cell death negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter animal organ development system development pronephros development brain morphogenesis stem cell differentiation positive regulation of epithelial cell proliferation mesenchymal to epithelial transition optic chiasma development cellular response to retinoic acid cellular response to glucose stimulus metanephric mesenchyme development positive regulation of mesenchymal to epithelial transition involved in metanephros morphogenesis metanephric mesenchymal cell differentiation nephric duct formation ureter development metanephric collecting duct development metanephric epithelium development metanephric distal convoluted tubule development metanephric nephron tubule formation positive regulation of metanephric glomerulus development negative regulation of mesenchymal cell apoptotic process involved in metanephric nephron morphogenesis regulation of metanephric nephron tubule epithelial cell differentiation cochlea development cochlea morphogenesis negative regulation of mesenchymal cell apoptotic process involved in metanephros development negative regulation of apoptotic process involved in metanephric collecting duct development negative regulation of apoptotic process involved in metanephric nephron tubule development negative regulation of reactive oxygen species metabolic process positive regulation of metanephric DCT cell differentiation positive regulation of optic nerve formation negative regulation of transcription from RNA polymerase II promoter metanephros development ureteric bud development kidney development transcription, DNA-templated transcription from RNA polymerase II promoter multicellular organism development positive regulation of cell proliferation regulation of apoptotic process regulation of programmed cell death negative regulation of cysteine-type endopeptidase activity involved in apoptotic process protein kinase B signaling negative regulation of cytolysis oxidation-reduction process paramesonephric duct development renal system development mesonephric tubule development mesonephric tubule formation mesonephric duct development reactive oxygen species metabolic process positive regulation of branching involved in ureteric bud morphogenesis aging pancreas development response to nutrient levels cellular response to hydrogen peroxide cellular response to mechanical stimulus cellular response to epidermal growth factor stimulus ureter morphogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline