Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRALWVLGLC CVLLTFGSVR ADDEVDVDGT VEEDLGKSRE GSRTDDEVVQ REEEAIQLDG LNASQIRELR EKSEKFAFQA EVNRMMKLII NSLYKNKEIF LRELISNASD ALDKIRLISL TDENALSGNE ELTVKIKCDK EKNLLHVTDT GVGMTREELV KNLGTIAKSG TSEFLNKMTE AQEDGQSTSE LIGQFGVGFY SAFLVADKVI VTSKHNNDTQ HIWESDSNEF SVIADPRGNT LGRGTTITLV LKEEASDYLE LDTIKNLVKK YSQFINFPIY VWSSKTETVE EPMEEEEAAK EEKEESDDEA AARRR
GRP94 is a 803 amino acid protein belonging to the heat shock protein 90 family. It acts as a molecular chaperone that functions in the processing and transport of secreted proteins. GRP94 and its N-terminal fragment stimulates CTL expansion and maturation of human monocyte-derived dendritic cells (MDDC). It plays a central role in innate as well as acquired immunity, maturation and chemotaxis of dendritic cells, Ab production, cross-priming, as a potential marker in breast cancer and is a peptide acceptor in endoplasmic reticulum and an accessory to peptide loading of MHC class I molecules. Expression of GRP94 suppressed A23187-induced apoptosis and stabilized calcium homeostasis. GRP94 is expressed in melanoma or liver metastases of colon carcinoma cells, human gastric carcinoma BGC-823 cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 94 kDa glucose-regulated protein; 98 kDa protein kinase; Endoplasmin; endothelial cell (HBMEC) glycoprotein; epididymis luminal protein 35; epididymis secretory sperm binding protein Li 125m; gp96 homolog; GRP-94; Heat shock protein 90 kDa beta member 1; heat shock protein 90kDa beta (Grp94), member 1; heat shock protein 90kDa beta family member 1; Heat shock protein family C member 4; PPK 98; stress-inducible tumor rejection antigen gp96; tumor rejection antigen (gp96) 1; Tumor rejection antigen 1; unnamed protein product
Gene Aliases: ECGP; GP96; GRP94; HEL-S-125m; HEL35; HSP90B1; HSPC4; ppk98; TRA1
UniProt ID: (Pig) Q29092, (Human) P14625
Entrez Gene ID: (Pig) 397191, (Human) 7184
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support