Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GFP Antibodies

          Invitrogen

          GFP Polyclonal Antibody, Alexa Fluor™ 488

          92 Published Figures
          516 References
          View all (144) GFP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite GFP Polyclonal Antibody, Alexa Fluor™ 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (92)
          GFP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GFP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 93

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          GFP Antibody (A-21311) in WB

          Western Blot was performed using Anti-GFP Polyclonal Antibody, Alexa Fluor 488 (Product # A-21311) by loading whole cell extracts of untransfected and transiently transfected HEK-293E lysates: untransfected, 40 µg (Lane 1), empty vector control, 40 µg (Lane 2), p65-GFP, 40 µg (Lane 3), His-p65-YFP, 40 µg (Lane 4), H3-mCherry, 40 µg (Lane 5) and 25 ng Positope (Lane 6) were electrophoresed using NuPAGE™ 4-12% Bis-Tris Protein Gel (Product # NP0322BOX). Resolved proteins were then transferred onto a nitrocellulose membrane (Product # IB23001... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          GFP Antibody in Western Blot (WB)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          GFP Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GFP Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GFP Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GFP Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          GFP Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          GFP Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)

          Product Details

          A-21311

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000
          View 17 publications 17 publications

          Immunohistochemistry (IHC)

          1:200-1:1,000
          View 314 publications 314 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 8 publications 8 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 24 publications 24 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          1-10 µg/mL
          View 104 publications 104 publications

          Flow Cytometry (Flow)

          -
          View 29 publications 29 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 11 publications 11 publications
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Human, Mouse, Tag

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The GFP was isolated directly from the jellyfish Aequorea victoria.

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          200 µg

          Purification

          IgG fraction

          Storage buffer

          PBS, pH 7.5

          Contains

          5mM sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_221477

          Product Specific Information

          To achieve appropriate staining results on formalin-fixed, paraffin embedded tissue sections, protease Type XIV (Actinase E) pretreatment is required. The recommended dilution and incubation for protease is 1-2 mg/mL at 37°C for 10 minutes. Other enzyme and Heat Induced Epitope Retrieval pre-treatment methods may not be suitable for this particular antibody. Frozen sections have been successfully stained after fixation with acetone at 4°C.

          Target Information

          The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: GFP; GFP tag; GFP2; green fluorescence; green fluorescent; green fluorescent protein; Turbo eGFP

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-26whj:80/100.66.74.142:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline