Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Secondary Antibodies ›
          • Donkey anti-Rabbit IgG Secondary Antibodies
          Catalog #: A32790 is a higher specification reagent that provides added control over experimental variables: Higher grade Alexa Fluor Plus Conjugates are engineered to provide more signal, lower cross-reactivity, and less low affinity interactions. Start seeing more signal and less noise today.

          Invitrogen

          Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488

          89 Published Figures
          View all (73) Donkey anti-Rabbit IgG antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Questions & Answers
          Tech Support

          Cite Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 488

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (10)
          • Published Figures (89)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 99

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody (A-21206) in ICC/IF

          Immunofluorescent analysis of PSD-95 (green) and MAP2 (red) on rat primary cortical neurons cultured for 28 days in the B-27 Plus Neuronal Culture System (Product # A3653401). At day 28 the cells were fixed with 4% paraformaldehyde for 15 min, permeabilized with 0.1% triton X-100 for 30min, and blocked with 1% BSA for 30 min at room temperature. Cells were stained with anti-PSD95 antibody (Product # 51-6900... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)
          Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          A-21206

          Applications
          Tested Dilution

          Immunohistochemistry (Paraffin) (IHC (P))

          1-10 µg/mL

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent

          Immunocytochemistry (ICC/IF)

          2 µg/mL

          Flow Cytometry (Flow)

          1-10 µg/mL
          Product Specifications

          Species Reactivity

          Rabbit

          Host/Isotype

          Donkey / IgG

          Class

          Polyclonal

          Type

          Secondary Antibody

          Immunogen

          Gamma Immunoglobins Heavy and Light chains

          Conjugate

          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Alexa Fluor 350 
          • Alexa Fluor 546 
          • Alexa Fluor 555 
          • Alexa Fluor 568 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 790 
          • Alexa Fluor Plus 405 
          • Alexa Fluor Plus 488 
          • Alexa Fluor Plus 555 
          • Alexa Fluor Plus 594 
          • Alexa Fluor Plus 647 
          • Alexa Fluor Plus 680 
          • Alexa Fluor Plus 800 
          • AP
          • Biotin 
          • FITC
          • HRP
          • TRITC
          • Unconjugated
          • Request custom conjugation

          Excitation/Emission Max

          499/520 nm View spectra spectra

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          1 mg

          Purification

          purified

          Storage buffer

          PBS, pH 7.5

          Contains

          5mM sodium azide

          Storage conditions

          4°C, store in dark

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2535792

          Target

          IgG

          Cross Adsorption

          Against bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat, and sheep serum

          Antibody Form

          Whole Antibody

          Product Specific Information

          To minimize cross-reactivity, these donkey anti-rabbit IgG whole antibodies have been affinity-purified and show minimum cross-reactivity to bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rat, and sheep serum proteins. Cross-adsorption or pre-adsorption is a purification step to increase specificity of the antibody resulting in higher sensitivity and less background staining. The secondary antibody solution is passed through a column matrix containing immobilized serum proteins from potentially cross-reactive species. Only the nonspecific-binding secondary antibodies are captured in the column, and the highly specific secondaries flow through. The benefits of this extra step are apparent in multiplexing/multicolor-staining experiments (e.g., flow cytometry) where there is potential cross-reactivity with other primary antibodies or in tissue/cell fluorescent staining experiments where there may be the presence of endogenous immunoglobulins.

          Alexa Fluor dyes are among the most trusted fluorescent dyes available today. Invitrogen™ Alexa Fluor 488 dye is a bright, green-fluorescent dye with excitation ideally suited to the 488 nm laser line. For stable signal generation in imaging and flow cytometry, Alexa Fluor 488 dye is pH-insensitive over a wide molar range. Probes with high fluorescence quantum yield and high photostability allow detection of low-abundance biological structures with great sensitivity. Alexa Fluor 488 dye molecules can be attached to proteins at high molar ratios without significant self-quenching, enabling brighter conjugates and more sensitive detection. The degree of labeling for each conjugate is typically 2-8 fluorophore molecules per IgG molecule; the exact degree of labeling is indicated on the certificate of analysis for each product lot.

          Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining. Because staining protocols vary with application, the appropriate dilution of antibody should be determined empirically. For the fluorophore-labeled antibodies a final concentration of 1-10 µg/mL should be satisfactory for most immunohistochemistry and flow cytometry applications.

          Product will be shipped at Room Temperature.

          Target Information

          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody A-21206 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline