Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Golgin-97 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          Golgin-97 Monoclonal Antibody (CDF4)

          Advanced Verification
          56 Published Figures
          218 References
          View all (19) Golgin-97 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support
          Datasheet
          Certificates
          Protocols
          Tech Support

          Cite Golgin-97 Monoclonal Antibody (CDF4)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (19)
          • Published Figures (56)
          • Advanced Verification (3)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 78

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Golgin-97 Antibody (A-21270) in ICC/IF

          Increased label specificity and resolution afforded by Image-iT® FX signal enhancer. Fixed and permeabilized MRC-5 human lung fibroblast cells were labeled with mouse anti-human Golgin-97 primary antibody (Product # A-21270), then visualized with either FITC-conjugated goat anti-mouse IgG (F2761, top row) or Alexa Fluor® 488 Goat Anti-Mouse IgG (Product # A-11001, bottom row). Cells on the right were treate... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry, Immunoprecipitation (ICC/IF, IP)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunocytochemistry (ICC/IF)
          Golgin-97 Antibody in Immunohistochemistry (IHC)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Western Blot (WB)
          Golgin-97 Antibody in Immunoprecipitation (IP)
          Golgin-97 Antibody in Immunoprecipitation (IP)
          Golgin-97 Antibody in Immunomicroscopy (IM)
          Golgin-97 Antibody
          Golgin-97 Antibody
          Golgin-97 Antibody

          Product Details

          A-21270

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1000
          View 26 publications 26 publications

          Immunohistochemistry (IHC)

          -
          View 10 publications 10 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100
          View 157 publications 157 publications

          Flow Cytometry (Flow)

          -
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 3 publications 3 publications

          Immunomicroscopy (IM)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 16 publications 16 publications
          Product Specifications

          Published species

          Chicken, Dog, Human, Monkey, Mouse, Non-human primate

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          CDF4

          Immunogen

          Human golgin-97.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Amount

          100 µg

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_221447

          Product Specific Information

          Anti-human golgin-97 antibodies were originally isolated from the serum of a patient with the autoimmune disease known as Sjögren's syndrome. These antibodies recognize a 97 kDa protein called golgin-97, a member of the granin family of proteins and a peripheral membrane protein localized on the cytoplasmic face of the Golgi apparatus. Because the antibody recognizes a protein unique to the Golgi apparatus of most vertebrate species, it is useful for immunodetection and identification of the Golgi apparatus in cells.

          To prepare a stock solution, reconstitute the antibody in 0.5-1 mL of phosphate-buffered saline, pH 7.4, containing 1% BSA. Store the solution for up to two weeks at 4°C with the addition of 2 mM sodium azide. For longer storage, divide the solution into single-use aliquots and freeze at -20°C.

          Target Information

          The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline