Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat
This gene contains long CAG trinucleotide repeats coding consecutive glutamine residues. The gene product may functions as a transcription factor, with a potential role in the regulation of neurodevelopment or neuroplasticity. The protein appears to bind and regulate the promoters of MMP1, MMP3, MMP7 and COL1A1. Studies in mouse suggest that nuclear matrix transcription factors (NP/NMP4) may be part of a general mechanical pathway that couples cell construction and function during extracellular matrix remodeling. Multiple transcript variants encoding several isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAG repeat protein 1; CAS-interacting zinc finger protein; expanded repeat domain, CAG/CTG 2; Nuclear matrix protein 4; Nuclear matrix transcription factor 4; Trinucleotide repeat-containing gene 1 protein; unnamed protein product; Zinc finger protein 384
Gene Aliases: CAGH1; CAGH1A; CIZ; ERDA2; NMP4; NP; TNRC1; ZNF384
UniProt ID: (Human) Q8TF68
Entrez Gene ID: (Human) 171017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support