Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • TAF1 Antibodies

          Bethyl Laboratories

          TAF1 Polyclonal Antibody

          View all (13) TAF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite TAF1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          TAF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TAF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TAF1 Antibody (A303-505A) in WB

          Detection of human and mouse TAF1 by western blot. Samples: Whole cell lysate (15 µg) from Jurkat, HeLa, HEK293T, TCMK-1, and NIH 3T3 cells prepared using NETN lysis buffer. Antibody: Affinity purified rabbit anti-TAF1 antibody (Product # A303-505A) (lot A303-505A-2) used for WB at 0.04 µg/mL. Detection: Chemiluminescence with an exposure time of 3 seconds. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TAF1 Antibody in Western Blot (WB)
          TAF1 Antibody in Immunoprecipitation (IP)
          TAF1 Polyclonal Antibody

          Product Details

          A303-505A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:2,000-1:10,000
          -

          Immunoprecipitation (IP)

          2-10 µg/mg lysate
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Region between residue 1800 and 1850 of Human Transcription Initiation Factor TFIID Subunit 1.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          tris citrate/phosphate, pH 7-8

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          The recommended shelf life for this product is 1 year from date of receipt.

          Based on 100% sequence identity, this antibody is predicted to react with Mouse and Golden hamster

          Target Information

          3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cell cycle gene 1 protein; cell cycle, G1 phase defect; complementation of cell cycle block, G1-to-S; p250; TAF(II)250; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, neuron specific isoform; TBP-associated factor 250 kDa; transcription factor TFIID p250 polypeptide; Transcription initiation factor TFIID 250 kDa subunit; Transcription initiation factor TFIID subunit 1

          View more View less

          Gene Aliases: AU015687; B430306D02Rik; BA2R; Ccg-1; CCG1; CCGS; DYT3; DYT3/TAF1; KAT4; MRXS33; N-TAF1; NSCL2; OF; P250; TAF(II)250; TAF1; TAF2A; TAFII-250; TAFII250; XDP

          View more View less

          UniProt ID: (Human) P21675, (Mouse) Q80UV9

          View more View less

          Entrez Gene ID: (Human) 6872, (Mouse) 270627

          View more View less

          Function(s)
          nucleotide binding RNA polymerase II core promoter sequence-specific DNA binding RNA polymerase II general transcription initiation factor binding transcription factor activity, core RNA polymerase I binding p53 binding DNA binding histone acetyltransferase activity protein kinase activity protein serine/threonine kinase activity protein binding ATP binding obsolete general RNA polymerase II transcription factor activity kinase activity transferase activity transferase activity, transferring acyl groups ligand-dependent nuclear receptor binding TBP-class protein binding sequence-specific DNA binding protein heterodimerization activity H3K27me3 modified histone binding RNA polymerase II sequence-specific DNA binding transcription factor binding ubiquitin conjugating enzyme activity peptide-lysine-N-acetyltransferase activity protein serine kinase activity histone H4K16ac reader activity transcription regulator inhibitor activity histone reader activity acetyl-CoA binding transcription coactivator activity transcription factor binding lysine-acetylated histone binding general transcription factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter protein polyubiquitination chromatin organization chromatin remodeling DNA-templated transcription, initiation transcription initiation from RNA polymerase I promoter transcription from RNA polymerase II promoter transcription initiation from RNA polymerase II promoter ubiquitin-dependent protein catabolic process cellular response to DNA damage stimulus negative regulation of gene expression midbrain development positive regulation of proteasomal ubiquitin-dependent protein catabolic process cellular response to UV mRNA transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter protein autophosphorylation protein stabilization RNA polymerase II transcriptional preinitiation complex assembly positive regulation of transcription initiation from RNA polymerase II promoter cellular response to ATP positive regulation of androgen receptor signaling pathway regulation of signal transduction by p53 class mediator negative regulation of signal transduction by p53 class mediator regulation of cell cycle G1/S phase transition negative regulation of protein autoubiquitination negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process transcription, DNA-templated regulation of transcription, DNA-templated cell cycle phosphorylation histone acetylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0