Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Orangutan
This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aldehyde dehydrogenase family 18 member A1; Delta-1-pyrroline-5-carboxylate synthase; delta-1-pyrroline-5-carboxylate synthetase; delta1-pyrroline-5-carboxlate synthetase; glutamate gamma-semialdehyde synthetase; MGC117316; P5CS; pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase); spastic paraplegia 9 (autosomal dominant); Spastic paraplegia-9 (spastic paraparesis with amyotrophy, cataracts and gastroesophageal reflux); unnamed protein product
Gene Aliases: 2810433K04Rik; ADCL3; AI429789; ALDH18A1; ARCL3A; GSAS; P5CS; PYCS; SPG9; SPG9A; SPG9B
UniProt ID: (Human) P54886, (Mouse) Q9Z110
Entrez Gene ID: (Human) 5832, (Mouse) 56454
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support