Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. tropicalis, Bovine and Pig
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-phosphate/5'-hydroxy nucleic acid ligase; ankyrin repeat domain 54; dJ149A16.6; FAAP; Focal adhesion-associated protein; HSPC117; RNA 2',3'-cyclic phosphate and 5'-OH ligase; RNA-splicing ligase RTCB; tRNA-splicing ligase RtcB homolog; unnamed protein product
Gene Aliases: AI255213; AI463255; C22orf28; D10Wsu52e; DJ149A16.6; FAAP; HSPC117; RTCB
UniProt ID: (Human) Q9Y3I0, (Mouse) Q99LF4
Entrez Gene ID: (Human) 51493, (Mouse) 28088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support