Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Dinitrophenyl-KLH Antibodies

          Invitrogen

          Dinitrophenyl-KLH Polyclonal Antibody

          7 Published Figures
          69 References
          View all (3) Dinitrophenyl-KLH antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Dinitrophenyl-KLH Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (7)
          Dinitrophenyl-KLH Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Dinitrophenyl-KLH Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 10

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Dinitrophenyl-KLH Antibody (A-6430) in ICC/IF

          A mixed population of trinitrophenyl-sensitized and nonsensitized bovine pulmonary artery endothelial cells that have been probed with anti-dinitrophenyl-KLH antibody (Product # A-6430), incubated with rabbit complement and then stained using the LIVE/DEAD® Reduced Biohazard Viability/Cytotoxicity Kit #1 (Product # L-7013). With this kit, the membrane-permeant SYTO® 10 dye labels the nucleic acids of live c... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Dinitrophenyl-KLH Antibody in Immunocytochemistry (ICC/IF)
          Dinitrophenyl-KLH Antibody in Immunohistochemistry (IHC)
          Dinitrophenyl-KLH Antibody in Immunohistochemistry (IHC)
          Dinitrophenyl-KLH Antibody in Immunohistochemistry (IHC)
          Dinitrophenyl-KLH Antibody in Immunohistochemistry (IHC)
          Dinitrophenyl-KLH Antibody in Western Blot (WB)
          Dinitrophenyl-KLH Antibody in Western Blot (WB)
          Dinitrophenyl-KLH Antibody in Western Blot (WB)
          Dinitrophenyl-KLH Antibody in Western Blot (WB)
          Dinitrophenyl-KLH Antibody in Cytotoxicity Assay (Cytotoxicity)

          Product Details

          A-6430

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 17 publications 17 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 10 publications 10 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 6 publications 6 publications

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 15 publications 15 publications

          Flow Cytometry (Flow)

          Assay-dependent
          View 2 publications 2 publications

          ELISA (ELISA)

          -
          View 4 publications 4 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 4 publications 4 publications

          Dot blot (DB)

          -
          View 1 publication 1 publication

          Cytotoxicity Assay (Cytotoxicity)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 8 publications 8 publications
          Product Specifications

          Species Reactivity

          Chemical

          Published species

          Chemical, Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Dinitrophenyl-KLH

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          1 mg

          Purification

          purified

          Storage buffer

          PBS, pH 7.2

          Contains

          5mM sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_221552

          Target Information

          DNP-KLH antibodies are used for detecting DNP-labeled molecules. Fluorescent and biotinylated anti-DNP-KLH conjugates can serve as second-step reagents for signal amplification in protein and nucleic acid detection systems, including in situ hybridization studies employing DNP-modified DNA probes. Because anti-DNP antibody binds well to DAMP (D-1552), an acidotropic reagent that can penetrate cell membranes, anti-DNP-KLH antibodies can also be used with DAMP for immunochemical localization of intracellular compartments with low pH. DNP-KLH antibodies may recognize compounds with a trinitrophenyl group, allowing the isolation of cell-surface molecules of human melanoma cells that have been labeled with trinitrobenzene sulfonic acid (TNBS).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C6H3N2O4; DNP-KLH

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline