Search
Search
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The testis spermatocyte apoptosis-related gene 2 protein (TSARG2, also designated spermatogenesis associated-4 or SPATA4) is involved in spermatogenesis. TSARG2 is specifically expressed in spermatogonia and spermatocytes of the seminiferous tubules, and it localizes to the nucleus. The predicted molecular weight of TSARG2 ranges depending on the species. TSARG2 is significantly upregulated in cryptorchidism and therefore, is a testis-specific apoptosis candidate oncogene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: spermatogenesis related gene 2; spermatogenesis-associated 4; Spermatogenesis-associated protein 4; Spermatogenesis-related gene 2 protein; testis and spermatogenesis cell related protein 2; Testis and spermatogenesis cell-related protein 2; Testis spermatocyte apoptosis-related gene 2 protein
Gene Aliases: 1700001N01Rik; CFAP178; FAP178; SPATA4; SPEF1B; Srg2; TSARG2
UniProt ID: (Human) Q8NEY3, (Mouse) Q8K3V1
Entrez Gene ID: (Human) 132851, (Mouse) 69281, (Rat) 306441
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support