Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QDEQVGLPGK GGKAKGKKTQ MAEVLPSPRG QRVIPRITIE MKAEAEKKNK KKIKNENTEG SPQEDGVELE GLKQRLEKKQ KREPGTKTKK QTTLAFKPIK KGKKRNPWSD SESDRSSDES NFDVPPRETE PRRAATKTKF TMDLDSDEDF SDFDEKTDDE DFVPSDASPP KTKTSPKLSN KELKPQKSVV SDLEADDVKG SVPLSSSPPA THFPDETEIT NPVPKKNVTV KKTAAKSQSS TSTTGAKKRA APKGTKRDPA LNSGVSQKPD PAKTKNRRKR KPSTSDDSDS NFEKIVSKAV TSKKSKGESD DFHMDFDSAV APRAKSVRAK KPIKYLEESD EDDLF
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2 and M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA gyrase; DNA topoisomerase (ATP-hydrolyzing); DNA topoisomerase 2-alpha; DNA topoisomerase II (EC 5.99.1.3); DNA Topoisomerase II alpha; DNA topoisomerase II, 170 kD; DNA topoisomerase II, alpha isozyme; EC 5.99.1.3; OTTHUMP00000207108; P11388-1; TOP2a/90; topoisomerase (DNA) II alpha 170kDa; topoisomerase ii alpha
Gene Aliases: Top-2; TOP2; TOP2A; TOP2alpha; TOPIIA; TP2A
UniProt ID: (Human) P11388, (Mouse) Q01320
Entrez Gene ID: (Human) 7153, (Mouse) 21973
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support