Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • SREBP1 Antibodies

          Proteintech

          SREBF1 Monoclonal Antibody (1B6G5), CoraLite® 594

          View all (19) SREBP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite SREBF1 Monoclonal Antibody (1B6G5), CoraLite® 594

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          SREBF1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SREBF1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SREBF1 Antibody (CL59466875100UL) in ICC/IF

          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite 594 SREBF1 antibody (Product # CL59466875100UL), Clone: 1B6G5 ) at dilution of 1:100. Green: stain with CoraLite 488-Phalloidin (CatNo. PF00001). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SREBF1 Antibody in Immunocytochemistry (ICC/IF)

          Product Details

          CL59466875100UL

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1B6G5

          Conjugate

          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation

          Excitation/Emission Max

          593/614 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles, store in dark

          Shipping conditions

          Wet ice

          Target Information

          Sterol regulatory element-binding proteins (SREBPs) are transcription factors that are members of the basic helix-loop-helix leucine zipper family of DNA binding proteins. Three isoforms have been identified in mammalian tissues that vary in structure, regulation, and function. SREBP-1a and SREBP-1c (originally cloned as ADD1) are protein products of alternative promoter usage of the SREBP-1 gene. The third isoform is transcribed from a different gene, SREBP-2. SREBPs are present as 120 kDa inactive precursors in the endoplasmic reticulum (ER) membrane. Upon activation, the SREBP protein is translocated to the Golgi and proteolytic cleavage occurs resulting in a mature transcriptionally active 60-78 kDa fragment. In liver and adipose tissues, SREBPs have a significant influence on lipid and cholesterol accumulation by inducing the transcription of genes involved in these processes. While SREBP-1 is thought to be more important in regulating the expression of genes involved in triglyceride synthesis and accumulation, SREBP-2 has been more closely linked to those involved in cholesterol synthesis and accumulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: bHLHd1; Class D basic helix-loop-helix protein 1; nt-SREBP1; SREBP-1; Sterol regulatory element-binding protein 1; Sterol regulatory element-binding transcription factor 1; unnamed protein product

          View more View less

          Gene Aliases: BHLHD1; HMD; IFAP2; SREBF1; SREBP1

          View more View less

          UniProt ID: (Human) P36956

          View more View less

          Entrez Gene ID: (Human) 6720

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription cofactor binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding chromatin binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding protein kinase binding sterol response element binding sequence-specific DNA binding macromolecular complex binding protein dimerization activity sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of heart rate by chemical signal regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter lipid metabolic process cholesterol biosynthetic process response to nutrient circadian rhythm steroid metabolic process cholesterol metabolic process insulin receptor signaling pathway lipid biosynthetic process cellular response to starvation response to xenobiotic stimulus response to glucose response to fructose positive regulation of triglyceride biosynthetic process regulation of fatty acid metabolic process insulin secretion lung development intracellular receptor signaling pathway regulation of protein stability response to food response to retinoic acid response to progesterone cellular response to insulin stimulus SREBP signaling pathway response to glucagon response to lipid mRNA transcription from RNA polymerase II promoter response to peptide hormone fat cell differentiation response to ethanol positive regulation of cholesterol biosynthetic process positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin secretion regulation of insulin secretion response to cAMP response to fatty acid cellular response to fatty acid negative regulation of triglyceride metabolic process regulation of mitophagy positive regulation of pri-miRNA transcription from RNA polymerase II promoter regulation of protein targeting to mitochondrion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline