Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • MAN2B2 Antibodies

          Abnova

          MAN2B2 Polyclonal Antibody, MaxPab™

          View all (4) MAN2B2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite MAN2B2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MAN2B2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAN2B2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAN2B2 Antibody (H00023324-B01P) in WB

          Western Blot analysis of MAN2B2 expression in transfected 293T cell line by MAN2B2 MaxPab polyclonal antibody. Lane 1: MAN2B2 transfected lysate(110.99 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAN2B2 Antibody in Western Blot (WB)

          Product Details

          H00023324-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAN2B2 (AAH94773.1, 1 a.a. approximately 1009 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGQLCWLPLL APLLLLRPPG VQSAGPIRAF VVPHSHMDVG WVYTVQESMR AYAANVYTSV VEELARGQQR RFIAVEQEFF RLWWDGVASD QQKYQVRQLL EEGRLEFVIG GQVMHDEAVT HLDDQILQLT EGHGFLYETF GIRPQFSWHV DPFGASATTP TLFALAGFNA HLGSRIDYDL KAAMQEARGL QFVWRGSPSL SERQEIFTHI MDQYSYCTPS HIPFSNRSGF YWNGVAVFPK PPPDGVYPNM SEPVTPANIN LYAEALVANV KQRAAWFRTP HVLWPWGCDK QFFNASVQFA NMDPLLDHIN SHAAELGVSV QYATLGDYFR ALHALNVTWR VRDHHDFLPY STEPFQAWTG FYTSRSSLKG LARRASALLY AGESMFTRYL WPAPRGHLDP TWALQQLQQL RWAVSEVQHH DAITGTESPK VRDMYATHLA SGMLGVRKLM ASIVLDELQP QAPMAASSDA GPAGHFASVY NPLAWTVTTI VTLTVGFPGV RVTDEAGHPV PSQIQNSTET PSAYDLLILT TIPGLSYRHY SIRPTAGAQE GTQEPAATVA STLQFGRRLR RRTSHAGRYL VPVANDCYIV LLDQDTNLMH SIWERQSNRT VRVTQEFLEY HVNRDVKQGP ISDNYLFTPG KAAVPAWEAV EMEIVAGQLV TEIRQYFYRN MTAQNYTYAI RSRLTHVPQG HDGELLCHRI EQEYQAGPLE LNREAVLRTS TNLNSQQVIY SDNNGYQMQR RPYVSYVNNS IARNYYPMVQ SAFMEDGKSR LVLLSERAHG ISSQGNGQVE VMLHRRLWNN FDWDLGYNLT LNDTSVVHPV LWLLLGSWSL TTALRQRSAL ALQHRPVVLF GDLAGTAPKL PGPQQQEAVT LPPNLHLQIL SIPGWRYSSN HTEHSQNLRK GHRGEAQADL RRVLLRLYHL YEVGEDPVLS QPVTVNLEAV LQALGSVVAV EERSLTGTWD LSMLHRWSWR TGPGRHRGDT TSPSRPPGGP IITVHPKEIR TFFIHFQQQ

          Target Information

          The MAN2B2 (Mannosidase Alpha Class 2B Member 2) gene encodes an enzyme that is involved in the processing and modification of glycoproteins by hydrolyzing alpha-mannosidic linkages. Located on chromosome 4, the MAN2B2 protein is crucial for proper lysosomal function and is part of the glycosyl hydrolase family. Pathogenic variants in the MAN2B2 gene have been linked to a rare congenital disorder of glycosylation (CDG), known as MAN2B2-CDG, characterized by immune deficiency, developmental delay, stroke, and dysmorphic facial features. These mutations lead to an altered mannosylation profile, impacting glycoprotein processing and cellular functions. Studies have shown that reintroducing wild-type MAN2B2 can restore the normal mannosylation profile in patient cells, highlighting its therapeutic potential. Furthermore, recent research has identified circular RNA from this gene (circMAN2B2) that is implicated in the proliferation of hepatocellular carcinoma cells, suggesting a role in cancer biology.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Alpha-1,6-mannosidase; Core-specific lysosomal alpha-1,6-Mannosidase; epididymis secretory sperm binding protein; Epididymis-specific alpha-mannosidase; Epman; HMG17L3; Mannosidase alpha class 2B member 2; NHC; unnamed protein product

          View more View less

          Gene Aliases: CDG1EE; EpMAN; KIAA0935; MAN2B2

          View more View less

          UniProt ID: (Human) Q9Y2E5

          View more View less

          Entrez Gene ID: (Human) 23324

          View more View less

          Function(s)
          catalytic activity alpha-mannosidase activity protein binding hydrolase activity hydrolase activity, acting on glycosyl bonds carbohydrate binding metal ion binding mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
          Process(es)
          carbohydrate metabolic process mannose metabolic process glycoprotein catabolic process oligosaccharide catabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0