Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ZFP95 Antibodies

          Abnova

          ZKSCAN5 Polyclonal Antibody, MaxPab™

          View all (6) ZFP95 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ZKSCAN5 Polyclonal Antibody, MaxPab™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          ZKSCAN5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ZKSCAN5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZKSCAN5 Antibody (H00023660-B01P) in WB

          Western Blot analysis of ZKSCAN5 expression in transfected 293T cell line by ZKSCAN5 MaxPab polyclonal antibody. Lane 1: ZFP95 transfected lysate(92.29 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZKSCAN5 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          H00023660-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ZKSCAN5 (AAH30790.1, 1 a.a. approximately 839 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.35 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MIMTESREVI DLDPPAETSQ EQEDLFIVKV EEEDCTWMQE YNPPTFETFY QRFRHFQYHE ASGPREALSQ LRVLCCEWLR PELHTKEQIL ELLVLEQFLT ILPEEFQPWV REHHPESGEE AVAVIENIQR ELEERRQQIV ACPDVLPRKM ATPGAVQESC SPHPLTVDTQ PEQAPQKPRL LEENALPVLQ VPSLPLKDSQ ELTASLLSTG SQKLVKIEEV ADVAVSFILE EWGHLDQSQK SLYRDDRKEN YGSITSMGYE SRDNMELIVK QISDDSESHW VAPEHTERSV PQDPDFAEVS DLKGMVQRWQ VNPTVGKSRQ NPSQKRDLDA ITDISPKQST HGERGHRCSD CGKFFLQASN FIQHRRIHTG EKPFKCGECG KSYNQRVHLT QHQRVHTGEK PYKCQVCGKA FRVSSHLVQH HSVHSGERPY GCNECGKNFG RHSHLIEHLK RHFREKSQRC SDKRSKNTKL SVKKKISEYS EADMELSGKT QRNVSQVQDF GEGCEFQGKL DRKQGIPMKE ILGQPSSKRM NYSEVPYVHK KSSTGERPHK CNECGKSFIQ SAHLIQHQRI HTGEKPFRCE ECGKSYNQRV HLTQHQRVHT GEKPYTCPLC GKAFRVRSHL VQHQSVHSGE RPFKCNECGK GFGRRSHLAG HLRLHSREKS HQCRECGEIF FQYVSLIEHQ VLHMGQKNEK NGICEEAYSW NLTVIEDKKI ELQEQPYQCD ICGKAFGYSS DLIQHYRTHT AEKPYQCDIC RENVGQCSHT KQHQKIYSST KSHQCHECGR GFTLKSHLNQ HQGIHTGEKP FQCKECGMNF SWSCSLFKHL RSHERTDPIN TLSVEGSLL

          Target Information

          Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. A member of the Krüppel C2H2-type zinc-finger protein family, ZKSCAN5, also known as ZFP95, is a 839 amino acid protein containing thirteen C2H2-type zinc fingers, one KRAB A domain and one SCAN box domain. ZKSCAN5 is located in the nucleus and is highly expressed in testis with lower levels of expression in liver, kidney, brain, striated muscle, lung and heart. Many KRAB A box-containing proteins have been shown to function as transcription repressors, suggesting a possible function of ZKSCAN5. As a result of alternative splicing, three transcripts of ZKSCAN5 exist, however one of those transcripts is specific to testis. The mouse homolog of ZKSCAN5 is differentially expressed in spermatogenesis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: unnamed protein product; zinc finger protein 95 homolog; zinc finger protein homologous to Zfp95 in mouse

          View more View less

          Gene Aliases: ZFP-95; ZFP95; ZNF914; ZSCAN37

          View more View less

          UniProt ID: (Human) Q9Y2L8

          View more View less

          Entrez Gene ID: (Human) 23660

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription factor activity, sequence-specific DNA binding protein binding zinc ion binding
          Process(es)
          regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0