Search
Search
Bethyl Laboratories
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, African elephant and Crab-eating macaque
ATF7IP is a multifunctional nuclear protein that associates with heterochromatin. It can act as a transcriptional coactivator or corepressor depending upon its binding partners.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activating transcription factor 7-interacting protein 1; ATF-interacting protein; ATF-IP; ATF7-interacting protein; ATFa-associated Modulator; FLJ10688; hAM; mAM; MBD1-containing chromatin-associated factor 1; P621; transcriptional mediator; unnamed protein product
Gene Aliases: 2610204M12Rik; AM; ATF-IP; ATF7IP; ATF7IP1; MCAF; MCAF1; p621
UniProt ID: (Human) Q6VMQ6, (Mouse) Q7TT18
Entrez Gene ID: (Human) 55729, (Mouse) 54343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support