Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody recognizes the PLMII epitope of AGR3 (AG3) and AGR2 (AG2) proteins (19-20 kDa); secreted cytoplasmic proteins which can serve as markers of carcinogenesis.
AGR2 (Anterior Gradient 2), also known as AG2 (hAG-2, HAG2 in human), or GOB-4, and AGR3 (Anterior Gradient 3), also known as AG3 (hAG-3, HAG3 in human), or BCMP11, are secreted cytoplasmic proteins which are involved in metastasis induction and p53 tumour supressor inhibition. They may serve as molecular markers and potential therapeutic targets for hormone-responsive breast tumours; AGR2 was reported also as a marker of other carcinomas. Xenopus homolog of these proteins is associated with anteroposterior fate determination during early development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AG-2; AG-3; AGR2 SV-C; Anterior gradient 3 homolog; anterior gradient homolog 2; anterior gradient homolog 3; Anterior gradient protein 2 homolog; Anterior gradient protein 3; anterior gradient protein 3 homolog; Breast cancer membrane protein 11; epididymis secretory protein Li 116; HPC8; protein disulfide isomerase family A, member 17; Protein disulfide isomerase family A, member 18; secreted cement gland homolog; Secreted cement gland protein XAG-2 homolog; unnamed protein product
Gene Aliases: AG-2; AG-3; AG2; AG3; AGR2; AGR3; BCMP11; GOB-4; HAG-2; hAG-3; HAG3; HEL-S-116; HPC8; PDIA17; PDIA18; RIFTD; UNQ515/PRO1030; UNQ642/PRO1272; XAG-2
UniProt ID: (Human) O95994, (Human) Q8TD06
Entrez Gene ID: (Human) 10551, (Human) 155465
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support