Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Calcium Sensing Receptor Antibodies

          Invitrogen

          Calcium Sensing Receptor Monoclonal Antibody (HL 1499)

          2 Published Figures
          2 References
          View all (24) Calcium Sensing Receptor antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Calcium Sensing Receptor Monoclonal Antibody (HL 1499)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (2)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calcium Sensing Receptor Antibody (MA1-16940) in WB

          Western blot analysis of Calcium Sensing Receptor in Hek293 lysate (0.5 mg/mL). Samples were incubated with Calcium Sensing Receptor monoclonal antibody (Product # MA1-16940). Specific band for Calcium Sensing Receptor. This experiment was performed under reducing conditions using the 12-230 kDa separation system. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Antibody in Flow Cytometry (Flow)
          Calcium Sensing Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calcium Sensing Receptor Antibody in Western Blot (WB)
          Calcium Sensing Receptor Monoclonal Antibody (HL 1499)

          Product Details

          MA1-16940

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:50-1:200
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          2.5 µg/mL
          -

          ELISA (ELISA)

          1:100-1:2,000
          -

          Immunoprecipitation (IP)

          5-10 µg
          -

          Radioimmune Assays (RIA)

          Assay-Dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rabbit, Rat

          Published species

          Mouse, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HL 1499

          Immunogen

          N-terminal region of human Calcium Sensing Receptor.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2071589

          Product Specific Information

          Suggested positive control: antigen standard for CASR (transient overexpression lysate), human kidney protein.

          Target Information

          Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Ca sensing receptor; calcium receptor|CaR; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; Extracellular calcium-sensing receptor; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; parathyroid Ca(2+)-sensing receptor 1; Parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1

          View more View less

          Gene Aliases: CAR; CASR; EIG8; FHH; FIH; GPRC2A; hCasR; HHC; HHC1; HYPOC1; NSHPT; PCAR1

          View more View less

          UniProt ID: (Human) P41180, (Rat) P48442, (Mouse) Q9QY96

          View more View less

          Entrez Gene ID: (Rabbit) 100341543, (Human) 846, (Rat) 24247, (Mouse) 12374

          View more View less

          Function(s)
          G-protein coupled receptor activity integrin binding calcium ion binding protein binding amino acid binding protein kinase binding identical protein binding protein homodimerization activity ion channel binding metal ion binding magnesium ion binding drug binding polyamine binding signal transducer activity G-protein coupled receptor
          Process(es)
          ossification response to ischemia detection of calcium ion cellular calcium ion homeostasis signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway JNK cascade chemosensory behavior positive regulation of cell proliferation anatomical structure morphogenesis response to metal ion positive regulation of gene expression positive regulation of insulin secretion bile acid secretion cellular response to hepatocyte growth factor stimulus vasodilation positive regulation of vasoconstriction branching morphogenesis of an epithelial tube positive regulation of positive chemotaxis response to calcium ion regulation of calcium ion transport fat pad development positive regulation of ERK1 and ERK2 cascade cellular response to vitamin D cellular response to glucose stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hypoxia response to fibroblast growth factor positive regulation of calcium ion import regulation of presynaptic membrane potential adenylate cyclase-activating G protein-coupled cAMP receptor signaling pathway positive regulation of NLRP3 inflammasome complex assembly cellular response to peptide chloride transmembrane transport calcium ion transport apoptotic process response to organic cyclic compound positive regulation of ATPase activity calcium ion import
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline