Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Agrin Antibodies

          Invitrogen

          Agrin Monoclonal Antibody (Agr 33)

          1 Reference
          View all (10) Agrin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Agrin Monoclonal Antibody (Agr 33)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Agrin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Agrin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Agrin Antibody (MA1-20378) in IHC

          Immunohistochemistry analysis of rat skeletal muscle probed with Agrin Monoclonal Antibody (Agr 33) (Product # MA1-20378) at 1:100. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Agrin Antibody in Immunohistochemistry (IHC)
          Agrin Monoclonal Antibody (Agr 33)

          Product Details

          MA1-20378

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          Agr 33

          Immunogen

          Recombinant rat agrin, C-terminal construct.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.2, with 0.1mM PMSF, 50% glycerol

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_557527

          Product Specific Information

          MA1-20378 detects agrin in mouse and rat samples.

          Target Information

          Agrin is a molecule that resides in the basal lamina of muscle cells and directs key events in post synaptic differentiation. Most notably, Agrin is responsible for the clustering of acetylcholine receptors (AChRs) on the cell surface and their localization to the neuromuscular junction. Several Agrin variants have been identified which arise from alternative mRNA splicings. Agrin splice forms having inserts at two sites in the carboxy terminus designated "y" and "z" display a high affinity for AChRs, while splice forms lacking these inserts associate with AChRs weakly. Muscle alpha-dystroglycan has been postulated to be the receptor for the clustering activity of agrin; however, this is a point of contention. Tyrosine phosphorylation has been implicated as a required early step in AChR aggregation. Interestingly, a unique receptor tyrosine kinase, designated MuSK, has been discovered that interacts with Agrin and is specifically localized to developing muscle.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Agrin; RP23-139J18.2

          View more View less

          Gene Aliases: AGR; Agrin; Agrn; nmf380

          View more View less

          UniProt ID: (Mouse) A2ASQ1, (Rat) P25304

          View more View less

          Entrez Gene ID: (Mouse) 11603, (Rat) 25592

          View more View less

          Function(s)
          dystroglycan binding calcium ion binding protein binding acetylcholine receptor regulator activity chondroitin sulfate binding BMP binding ATPase inhibitor activity heparan sulfate proteoglycan binding ion channel binding transforming growth factor beta binding sialic acid binding protease inhibitor
          Process(es)
          regulation of protein phosphorylation positive regulation of protein phosphorylation plasma membrane organization chemical synaptic transmission multicellular organism development synapse assembly neuromuscular junction development regulation of synaptic growth at neuromuscular junction regulation of receptor activity cell differentiation positive regulation of protein binding regulation of GTPase activity receptor clustering positive regulation of neuron apoptotic process positive regulation of GTPase activity neurotransmitter receptor metabolic process positive regulation of synaptic growth at neuromuscular junction positive regulation of transcription from RNA polymerase II promoter protein heterotetramerization positive regulation of filopodium assembly regulation of cardiac muscle contraction positive regulation of protein tyrosine kinase activity regulation of microtubule cytoskeleton organization skeletal muscle acetylcholine-gated channel clustering regulation of cardiac muscle cell membrane potential regulation of axon guidance negative regulation of sodium ion export from cell negative regulation of sodium:potassium-exchanging ATPase activity positive regulation of protein geranylgeranylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline