Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Calprotectin Antibodies

          Invitrogen

          Calprotectin Monoclonal Antibody (MAC387)

          Advanced Verification
          7 Published Figures
          31 References
          View all (99) Calprotectin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Calprotectin Monoclonal Antibody (MAC387)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (7)
          • Advanced Verification (9)
          Calprotectin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Calprotectin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calprotectin Antibody (MA1-80155) in ICC/IF

          Immunofluorescence analysis of Calprotectin (S100A9) was performed using 70% confluent log phase THP-1 and Raji cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with Calprotectin (S100A9) Monoclonal Antibody (MAC387) (Product # MA1-80155) at 1:250 dilution ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calprotectin Antibody in Immunocytochemistry (ICC/IF)
          Calprotectin Antibody in Immunocytochemistry (ICC/IF)
          Calprotectin Antibody in Immunocytochemistry (ICC/IF)
          Calprotectin Antibody in Western Blot (WB)
          Calprotectin Antibody in Western Blot (WB)
          Calprotectin Antibody in Western Blot (WB)
          Calprotectin Antibody in Immunocytochemistry (ICC/IF)
          Calprotectin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calprotectin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calprotectin Antibody in Immunohistochemistry (IHC)
          Calprotectin Antibody in Immunohistochemistry (IHC)
          Calprotectin Antibody in Immunohistochemistry (IHC)
          Calprotectin Antibody in Immunohistochemistry (IHC)
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Antibody
          Calprotectin Monoclonal Antibody (MAC387)

          Product Details

          MA1-80155

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          -
          View 19 publications 19 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:200
          View 5 publications 5 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:250
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          1:50-1:100
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Bovine, Dog, Deer, Horse, Cat, Guinea pig, Goat, Human, Non-human primate, Pig, Rabbit, Rat

          Published species

          Bovine, Cynomolgus monkey, Dog, Horse, Human, Mouse, Pig, Rat, Rhesus monkey

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MAC387

          Immunogen

          Human monocytes.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • APC 
          • FITC
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          20 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2184278

          Product Specific Information

          This clone recognizes the L1 or Calprotectin molecule, an intracytoplasmic antigen comprised of a 12 kDa alpha chain and a 14 kDa beta chain. Although originally described as binding to epitopes common to both the alpha and beta chains (Flavell et al. 1987) subsequent studies indicate that the antibody detects an epitope exclusively expressed on the beta chain (Goebeler et al. 1994) demonstrated by immunofluorescent and western blotting on both naturally expressing and transfected targets. In addition Mouse anti Human macrophages, clone MAC387 detects the beta chain in complex with the alpha. The antigen recognized by Mouse anti Human macrophages, clone MAC387 is expressed by granulocytes, monocytes and by tissue macrophages. Variable results have been reported for staining brain macrophages and microglia. The epitope recognized appears to be well conserved and the antibody is routinely used for the detection of myeloid cellds in a wide range of species.

          This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase prior to IHC staining. A suggested positive control is human spleen. For flow cytometry, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.

          Target Information

          The S100A8/A9 heterocomplex, also called MRP-8/MRP-14 complex or calprotectin, comprises 60% of the cytoplasmic protein fraction of circulating polymorphonuclear granulocytes and is also found in monocytes, macrophages and ileal tissue eosinophils. Peripheral blood monocytes carry the antigen extra- and intracellularly, neutrophils only intracellularly. The S100A8/A9 complex has antibacterial, antifungal, immunomodulating and antiproliferative effects. Besides this it is a potent chemotactic factor for neutrophils. Plasma concentrations are elevated in diseases associated with increased neutrophil activity, like inflammatory bowel disease. Granulocytes terminate their existence after transmigration through the intestinal wall. Therefore calprotectin is also detectable in feces. Elevated levels of calprotectin have been observed in body fluids such as plasma, saliva, gingival crevicular fluid, stools, and synovial fluid during infection and inflammatory conditions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: AI323541; B8Ag; BEE11; BEE22; calcium binding protein; S100 protein; calgranulin A; calgranulin B; Calgranulin-A; Calgranulin-B; calprotectin; Calprotectin L1H subunit; Calprotectin L1L subunit; CFAG; Cystic fibrosis antigen; intracellular calcium-binding protein (MRP14); Leukocyte L1 complex heavy chain; Leukocyte L1 complex light chain; macrophage migration inhibitory factor-related protein-14; macrophage migration inhibitory factor-related protein-8; Migration inhibitory factor-related protein 14; Migration inhibitory factor-related protein 8; MRP-14; MRP-8; Myeloid-related protein 14; Neutrophil cytosolic 23 kDa protein; Neutrophil cytosolic 7 kDa protein; p14; p23; P7; p8; Protein S100-A8; Protein S100-A9; S100 calcium binding protein A8; S100 calcium binding protein A8 (calgranulin A); S100 calcium binding protein A9 (calgranulin B); S100 calcium-binding protein A8; S100 calcium-binding protein A8 (calgranulin A); S100 calcium-binding protein A9; S100 calcium-binding protein A9 (calgranulin B); S100A8; S100A8/S100A9; S100A9; unnamed protein product; Urinary stone protein band A

          View more View less

          Gene Aliases: 60B8AG; CAGA; CAGB; CFAG; CGLA; CGLB; CP-10; L1AG; LIAG; MA387; MAC387; MIF; MRP-14; MRP-8; MRP14; MRP8; NIF; P14; P8; S100-A8; S100-A9; S100A8; S100A9

          View more View less

          UniProt ID: (Human) P05109, (Bovine) P28782, (Bovine) P28783, (Human) P06702, (Rabbit) P50117, (Rat) P50115, (Rat) P50116

          View more View less

          Entrez Gene ID: (Guinea pig) 100735514, (Horse) 100061766, (Dog) 490461, (Human) 6279, (Bovine) 616818, (Pig) 100127488, (Rabbit) 100008985, (Goat) 102168876, (Dog) 490463, (Bovine) 532569, (Pig) 100127489, (Human) 6280, (Horse) 100061730, (Rabbit) 100008704, (Goat) 102182273, (Rat) 116547, (Guinea pig) 100735238, (Rat) 94195

          View more View less

          Function(s)
          calcium ion binding protein binding microtubule binding zinc ion binding Toll-like receptor 4 binding metal ion binding calcium-dependent protein binding arachidonic acid binding RAGE receptor binding antioxidant activity molecular_function calmodulin-related
          Process(es)
          immune system process leukocyte migration involved in inflammatory response chronic inflammatory response peptide secretion positive regulation of peptide secretion autophagy apoptotic process chemotaxis inflammatory response response to zinc ion astrocyte development peptidyl-cysteine S-nitrosylation positive regulation of cell growth neutrophil chemotaxis response to lipopolysaccharide regulation of toll-like receptor signaling pathway autocrine signaling defense response to bacterium endothelial cell migration innate immune response response to ethanol positive regulation of inflammatory response defense response to fungus regulation of cytoskeleton organization neutrophil aggregation positive regulation of intrinsic apoptotic signaling pathway activation of cysteine-type endopeptidase activity involved in apoptotic process cellular oxidant detoxification cell-cell signaling positive regulation of neuron projection development peptidyl-cysteine S-trans-nitrosylation regulation of respiratory burst involved in inflammatory response antimicrobial humoral immune response mediated by antimicrobial peptide acute inflammatory response wound healing leukocyte chemotaxis actin cytoskeleton reorganization regulation of integrin biosynthetic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline