Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • CD3 Antibodies

          Invitrogen

          CD3e Monoclonal Antibody (SP7)

          11 Published Figures
          290 References
          View all (474) CD3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD3e Monoclonal Antibody (SP7)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (11)
          CD3e Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CD3e Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 14

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD3e Antibody (MA1-90582) in IHC (P)

          Immunohistochemical analysis of CD3e in formalin-fixed, paraffin-embedded human tonsil tissue using a CD3E monoclonal antibody (Product # MA1-90582). Detection was performed with a peroxidase-conjugate and AEC chromagen. Note cell membrane staining of T cells. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD3e Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD3e Antibody in Western Blot (WB)
          CD3e Antibody in Flow Cytometry (Flow)
          CD3e Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD3e Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD3e Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CD3e Antibody in Immunohistochemistry (Frozen) (IHC (F))
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Antibody in Immunohistochemistry (IHC)
          CD3e Monoclonal Antibody (SP7)

          Product Details

          MA1-90582

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 247 publications 247 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150
          View 28 publications 28 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          -
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          1:1,000
          View 4 publications 4 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Baboon, Dog, Human, Mouse, Non-human primate, Pig, Rat, Rhesus monkey

          Host/Isotype

          Rabbit / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          SP7

          Immunogen

          synthetic peptide KAKAKPVTRGAGA, corresponding to amino acids 156-168 of Human CD3 epsilon chain.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.01 mg/mL

          Amount

          5 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2-7.4, with 1% BSA

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1956722

          Product Specific Information

          This antibody reacts with the intracytoplasmic portion of CD3 and stains human T cells in both the cortex and medula of the thymus and in peripheral lymphoid tissues. This antibody is suitable for staining normal and neoplastic T cells in formalin-fixed, paraffin embedded tissues. For tissue staining, antigen retrieval is suggested by boiling tissue sections in 10mM citrate buffer, pH 6.0 for 10 minutes followed by cooling at room temperature for 20 minutes.

          This antibody is predicted to react with Sheep, Rabbit, Horse, Chicken, Cow, Cat, Dog, Baboon, and Woodchuck.

          Recommended positive controls:

          WB - Recombinant human CD3e protein, Jurkat whole cell lysate, Human, mouse and rat thymus tissue lysate|

          IHC (P) - Pig and rat spleen tissue, human tonsil tissue, mouse epidydimal fat pad and lymph node tissues, rat infarcted heart and spleen tissues

          Flow Cytometry - Human peripheral blood lymphocytes, Jurkat cells

          IHC (F) - Mouse brain tissue

          Target Information

          The CD3 complex, composed of gamma, delta, epsilon, and zeta subunits, is essential for the assembly, trafficking, and surface expression of the T cell receptor (TCR) complex. These subunits are structurally related members of the immunoglobulin superfamily and are encoded by closely linked genes on human chromosome 11. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells, but not on B or NK cells. The CD3 subunits play a crucial role in transducing antigen-recognition signals into the cytoplasm of T cells. The cytoplasmic tails of CD3 subunits contain a double tyrosine-based motif that associates with cytoplasmic signal transduction molecules, mediating T cell activation through the TCR. Crosslinking of the TCR initiates intracellular biochemical pathways that result in cellular activation, proliferation, and potentially growth arrest and cell survival. CD3 is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes, and Purkinje cells in the cerebellum. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. Defects in the CD3 gene are associated with CD3 immunodeficiency, highlighting its importance in immune function and regulation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CD247; CD247 antigen; CD247 antigen, zeta subunit; CD3 antigen delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 gamma-chain; CD3 molecule delta polypeptide; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma polypeptide; CD3 TCR complex; CD3-epsilon; CD3-zeta/eta; CD3d; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule, delta (CD3-TCR complex); CD3e; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule, epsilon (CD3-TCR complex); CD3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3Z antigen, zeta polypeptide (TiT3 complex); CD3zeta chain; component of the TCR/CD3 complex; FLJ18683; Leu-4; OKT3, delta chain; T cell antigen receptor complex epsilon subunit of T3; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; T3/TCR complex; TCR zeta; TCR zeta chain; tissue factor-targeting CAR1 dimer; tissue factor-targeting CAR1 monomer; unnamed protein product

          View more View less

          Gene Aliases: 4930549J05Rik; A430104F18Rik; AI504783; AW552088; CD247; CD3; CD3-DELTA; Cd3-eta; CD3-GAMMA; CD3-ZETA; CD3D; CD3DELTA; CD3E; CD3epsilon; CD3G; CD3GAMMA; CD3H; CD3Q; CD3Z; CD3ZETA; Ctg-3; Ctg3; IMD17; IMD18; IMD19; IMD25; T3D; T3E; T3G; T3Z; TCRE; Tcrk; TCRZ; TCRzeta

          View more View less

          UniProt ID: (Human) P20963, (Human) P04234, (Human) P07766, (Human) P09693, (Mouse) P24161, (Mouse) P04235, (Rat) P19377, (Mouse) P22646, (Mouse) P11942, (Rat) Q64159

          View more View less

          Entrez Gene ID: (Human) 919, (Human) 915, (Human) 916, (Human) 917, (Rat) 25300, (Mouse) 12503, (Mouse) 12500, (Rat) 25710, (Rat) 315609, (Mouse) 12501, (Mouse) 12502, (Rat) 300678

          View more View less

          Function(s)
          transmembrane signaling receptor activity protein binding identical protein binding protein homodimerization activity protein heterodimerization activity protein tyrosine kinase binding SH3 domain binding protein kinase binding receptor signaling complex scaffold activity T cell receptor binding molecular_function transcription coactivator activity signal transducer activity defense/immunity protein immunoglobulin receptor superfamily
          Process(es)
          adaptive immune response immune system process cell surface receptor signaling pathway Fc-gamma receptor signaling pathway gamma-delta T cell activation alpha-beta T cell activation T cell receptor signaling pathway protein oligomerization protein-containing complex assembly positive regulation of protein localization to cell surface positive thymic T cell selection positive regulation of cell-matrix adhesion positive regulation of T cell anergy T cell anergy transmembrane receptor protein tyrosine kinase signaling pathway signal complex assembly G-protein coupled receptor signaling pathway smoothened signaling pathway regulation of signal transduction positive regulation of gene expression negative regulation of gene expression dendrite development calcium-mediated signaling cerebellum development T cell costimulation positive regulation of interferon-gamma production positive regulation of interleukin-2 production positive regulation of interleukin-4 production positive regulation of cell-cell adhesion mediated by integrin CD4-positive, alpha-beta T cell proliferation T cell proliferation positive regulation of T cell proliferation T cell activation regulation of apoptotic process positive regulation of MAPK cascade negative thymic T cell selection negative regulation of smoothened signaling pathway lymphocyte activation positive regulation of calcium-mediated signaling positive regulation of T cell activation apoptotic signaling pathway positive regulation of CD4-positive, alpha-beta T cell proliferation establishment or maintenance of cell polarity protein transport regulation of lymphocyte apoptotic process protein complex assembly biological_process T cell differentiation positive regulation of transcription from RNA polymerase II promoter response to nutrient T cell differentiation in thymus positive regulation of interleukin-2 biosynthetic process positive regulation of alpha-beta T cell proliferation positive regulation of peptidyl-tyrosine phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline