Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody recognizes heavy chain human skeletal myosin (fast-twitch type II isomyosin). It does not stain cardiac or smooth muscle myosin.
For antigen retrieval, it is recommended to use a citrate buffer, pH 6.0, at room temperature for 30 min.
Store as a concentrated solution. Centrifuge briefly prior to opening vial. Store at 4°C.
Myosin is a major contractile protein which converts chemical energy into mechanical energy through the hydrolysis of ATP. Myosin is a hexameric protein composed of a pair of myosin heavy chains (MYH) and two pairs of nonidentical light chains. Myosin heavy chains are encoded by a multigene family. In mammals at least 10 different myosin heavy chain (MYH) isoforms have been described from striated, smooth, and nonmuscle cells. These isoforms show expression that is spatially and temporally regulated during development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2A myosin heavy chain; 2B myosin heavy chain; epididymis luminal protein 71; fast 2a myosin heavy chain; inclusion body myopathy 3, autosomal dominant; MGC133384; myh4; MyHC-2a; MyHC-2b; MyHC-2x; MyHC-IIa; MyHC-IIb; MyHC-IIx/d; Myosin heavy chain 1; Myosin heavy chain 2; Myosin heavy chain 2a; Myosin heavy chain 2b; Myosin heavy chain 2x; Myosin heavy chain 4; Myosin heavy chain IIa; Myosin heavy chain IIb; Myosin heavy chain IIx/d; myosin heavy chain type IIb; myosin heavy chain type IIx; Myosin heavy chain, skeletal muscle; Myosin heavy chain, skeletal muscle, adult 1; Myosin heavy chain, skeletal muscle, adult 2; Myosin heavy chain, skeletal muscle, fetal; Myosin heavy chain, skeletal muscle, juvenile; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin, heavy polypeptide 4, skeletal muscle; Myosin-1; Myosin-2; Myosin-4; type 2X myosin heavy chain; type IIA myosin heavy chain; unnamed protein product
Gene Aliases: CMYO6; CMYP6; HEL71; IBM3; MHC; MYH1; MYH2; MYH2A; MYH2B; MYH4; MYHa; MYHas8; MYHC; MyHC-2A; MyHC-2B; MyHC-2x; MyHC-2X/D; MyHC-IIA; MyHC-IIb; MYHSA1; MYHSA2; MYPOP
UniProt ID: (Human) P12882, (Human) Q9UKX2, (Human) Q9Y623, (Rabbit) Q28641, (Rat) Q29RW1
Entrez Gene ID: (Human) 4619, (Human) 4620, (Human) 4622, (Rabbit) 100009548, (Rabbit) 100125991, (Rat) 287408, (Rat) 691644, (Rat) 360543
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support