Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ARF Pan Antibodies

          Invitrogen

          ARF1/ARF3/ARF5/ARF6 Monoclonal Antibody (1D9)

          Advanced Verification
          1 Published Figure
          20 References
          View all (3) ARF Pan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ARF1/ARF3/ARF5/ARF6 Monoclonal Antibody (1D9)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (13)
          • Published Figures (1)
          • Advanced Verification (1)
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ARF1/ARF3/ARF5/ARF6 Antibody (MA3-060) in ICC/IF

          Immunofluorescent analysis of ADP-Ribosylation Factor using ADP-Ribosylation Factor Monoclonal Antibody (1D9) (Product # MA3-060) shows staining in Hela Cells. ADP-Ribosylation Factor (green), F-Actin staining with Phalloidin (red) and nuclei with DAPI (blue) is shown. Cells were grown on chamber slides and fixed with formaldehyde prior to staining. Cells were probed without (control) or with an antibody re... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
          ARF1/ARF3/ARF5/ARF6 Antibody in Flow Cytometry (Flow)
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunocytochemistry (ICC/IF)
          ARF1/ARF3/ARF5/ARF6 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
          ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
          ARF1/ARF3/ARF5/ARF6 Antibody in Flow Cytometry (Flow)
          ARF1/ARF3/ARF5/ARF6 Antibody in Flow Cytometry (Flow)
          ARF1/ARF3/ARF5/ARF6 Antibody in Western Blot (WB)
          ARF1/ARF3/ARF5/ARF6 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          MA3-060

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:3000
          View 12 publications 12 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100
          -

          Immunocytochemistry (ICC/IF)

          1:100
          View 5 publications 5 publications

          Flow Cytometry (Flow)

          1/20
          View 1 publication 1 publication

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Hamster, Human, Mouse, Rat, Dog

          Published species

          Bovine, Hamster, Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1D9

          Immunogen

          Recombinant human ARF1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Storage buffer

          ascites

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2274419

          Product Specific Information

          MA3-060 detects ADP-ribosylation factor1 (Arf1), Arf3, Arf5 and Arf6 and about ten-fold less well with Arf4 from human, mouse and rat tissues.

          MA3-060 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, and immunoprecipitation procedures. By Western blot, this antibody detects a single ~21 kDa protein representing Arf in rat pancreas extract. Immunofluorescence staining of Arf in rat pancreas with MA3-060 results in staining of the cytoplasmic face of the trans-Golgi membranes. MA3-060 cannot be used to neutralize Arf in solution.

          The MA3-060 antigen is recombinant human Arf1.

          Target Information

          ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq, Jul 2008]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ADP ribosylation factor 1; ADP ribosylation factor 3; ADP ribosylation factor 4; ADP ribosylation factor 5; ADP ribosylation factor 6; ADP-ribosylation factor (ARF1); ADP-ribosylation factor 1; ADP-ribosylation factor 2; ADP-ribosylation factor 3; ADP-ribosylation factor 4; ADP-ribosylation factor 4-like protein; ADP-ribosylation factor 5; ADP-ribosylation factor 6; ARF; ARF6; liver regeneration-related protein LRRG202; pan ARF; small GTP binding protein; unnamed protein product

          View more View less

          Gene Aliases: 5430400P17Rik; Ac1-253; ARF2; I79_003112; PVNH8

          View more View less

          UniProt ID: (Human) P84077, (Human) P61204, (Human) P18085, (Human) P84085, (Human) P62330, (Rat) P84079, (Mouse) P84078, (Mouse) Q8BSL7, (Rat) P84082, (Mouse) P61205, (Rat) P61206, (Mouse) P61750, (Rat) P61751, (Mouse) P84084, (Rat) P84083, (Mouse) P62331, (Rat) P62332

          View more View less

          Entrez Gene ID: (Dog) 100856517, (Human) 375, (Human) 377, (Dog) 486562, (Dog) 607073, (Human) 378, (Human) 381, (Dog) 475206, (Dog) 490679, (Human) 382, (Chinese hamster) 100757864, (Rat) 64310, (Mouse) 11840, (Mouse) 11841, (Rat) 79119, (Chinese hamster) 100766631, (Mouse) 11842, (Rat) 140940, (Mouse) 11843, (Chinese hamster) 100769922, (Rat) 79120, (Mouse) 11844, (Rat) 79117, (Chinese hamster) 100759676, (Chinese hamster) 100774654, (Mouse) 11845, (Rat) 79121

          View more View less

          Function(s)
          magnesium ion binding RNA binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding protein domain specific binding guanyl-nucleotide exchange factor activity epidermal growth factor receptor binding NAD+-protein-arginine ADP-ribosyltransferase activity phospholipase D activator activity GDP binding protein binding, bridging thioesterase binding signaling adaptor activity G-protein
          Process(es)
          regulation of receptor internalization cellular copper ion homeostasis intracellular protein transport vesicle-mediated transport regulation of Arp2/3 complex-mediated actin nucleation glycolipid transport long term synaptic depression dendritic spine organization cellular response to virus protein localization to Golgi membrane mitotic cleavage furrow ingression retrograde vesicle-mediated transport, Golgi to ER ER to Golgi vesicle-mediated transport epidermal growth factor receptor signaling pathway cell migration negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter response to axon injury regulation of synapse organization cilium morphogenesis dendritic spine development regulation of postsynapse organization regulation of cilium assembly regulation of reactive oxygen species metabolic process positive regulation of TOR signaling lysosomal membrane organization cell adhesion regulation of neuron projection development positive regulation of neuron projection development positive regulation of actin filament polymerization cortical actin cytoskeleton organization endocytic recycling protein localization to cell surface regulation of Rac protein signal transduction protein localization to endosome negative regulation of receptor-mediated endocytosis synaptic vesicle endocytosis positive regulation of protein secretion regulation of filopodium assembly positive regulation of keratinocyte migration regulation of dendritic spine development protein localization to plasma membrane ruffle assembly maintenance of postsynaptic density structure positive regulation of focal adhesion disassembly positive regulation of protein localization to plasma membrane positive regulation of mitotic cytokinetic process protein localization to cleavage furrow regulation of presynapse assembly cellular response to nerve growth factor stimulus negative regulation of protein localization to cell surface negative regulation of dendrite development Golgi to plasma membrane transport actin filament organization very-low-density lipoprotein particle assembly positive regulation of endocytosis positive regulation of calcium ion-dependent exocytosis positive regulation of dendritic spine development synaptic vesicle budding postsynaptic actin cytoskeleton organization positive regulation of sodium ion transmembrane transport positive regulation of late endosome to lysosome transport positive regulation of ER to Golgi vesicle-mediated transport regulation of phospholipid metabolic process protein transport learning apical protein localization establishment or maintenance of epithelial cell apical/basal polarity protein localization to cilium liver development myeloid cell apoptotic process regulation of toll-like receptor 4 signaling pathway establishment of epithelial cell polarity hepatocyte apoptotic process actin cytoskeleton organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-d6wk2:80/100.66.72.150:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0