Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 25
MA3-911 detects sarcoplasmic or endoplasmic reticulum calcium 1 (SERCA1) ATPase from canine, human, rabbit, rat, mouse and guinea pig tissues.
MA3-911 has been successfully used in Western blot, immunohistochemistry, and immunofluorescence procedures. By Western blot, this antibody detects an ~110 kDa protein representing SERCA1 ATPase in canine skeletal muscle extracts. Immunofluorescence staining of SERCA1 ATPase in canine skeletal muscle with MA3-911 results in strong labeling of the entire type II (fast) myofiber.
The MA3-911 antigen is purified rabbit skeletal muscle sarcoplasmic reticulum. This antibody recognizes an epitope between amino acids 199-505 of rabbit skeletal muscle ATPase, a region that is not exposed in native sarcoplasmic reticulum.
ATP dependent calcium pumps are responsible, in part, for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium (SERCA) ATPases. The SERCA1 gene is exclusively expressed in type II (fast) skeletal muscle. The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific form expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. The SERCA3 gene is not as well characterized and is found in non-muscle cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: adult Ca2+ ATPase; ATPase, Ca++ transporting, cardiac muscle, fast twitch 1; ATPase, Ca++ transporting, ubiquitous; Ca2+ ATPase; Calcium pump 1; Calcium-transporting ATPase sarcoplasmic reticulum type, fast twitch skeletal muscle isoform; Endoplasmic reticulum class 1/2 Ca(2+) ATPase; neonatal Ca2+ ATPase; Sarcoplasmic/endoplasmic reticulum calcium ATPase 1; SERCA; SERCA ATPase; SERCA1; SR Ca(2+)-ATPase 1; unnamed protein product
Gene Aliases: ATP2A; ATP2A1; ATP2A3; SERCA1; SERCA1a
UniProt ID: (Human) O14983, (Rabbit) P04191, (Mouse) Q8R429, (Rat) Q64578
Entrez Gene ID: (Dog) 479797, (Human) 487, (Rabbit) 100037716, (Mouse) 11937, (Rat) 116601, (Guinea pig) 100726234
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support