Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Phospholamban Antibodies

          Invitrogen

          Phospholamban Monoclonal Antibody (2D12)

          Advanced Verification
          61 Published Figures
          180 References
          View all (28) Phospholamban antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Phospholamban Monoclonal Antibody (2D12)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (6)
          • Published Figures (61)
          • Advanced Verification (3)
          Phospholamban Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospholamban Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 70

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospholamban Antibody (MA3-922) in ICC/IF

          Immunofluorescent analysis of Phospholamban (green) showing staining in the cytoplasm of C2C12 cells (right) compared to a negative control without primary antibody (left). Formalin-fixed cells were permeabilized with 0.1% Triton X-100 in TBS for 5-10 minutes and blocked with 3% BSA-PBS for 30 minutes at room temperature. Cells were probed with a Phospholamban monoclonal antibody (Product # MA3-922) in 3% B... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospholamban Antibody in Immunocytochemistry (ICC/IF)
          Phospholamban Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospholamban Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospholamban Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospholamban Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospholamban Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospholamban Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospholamban Antibody in Immunohistochemistry (IHC)
          Phospholamban Antibody in Immunohistochemistry (IHC)
          Phospholamban Antibody in Immunohistochemistry (IHC)
          Phospholamban Antibody in Immunohistochemistry (IHC)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Western Blot (WB)
          Phospholamban Antibody in Immunoprecipitation (IP)
          Phospholamban Antibody
          Phospholamban Antibody
          Phospholamban Antibody
          Phospholamban Monoclonal Antibody (2D12)

          Product Details

          MA3-922

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:5,000
          View 162 publications 162 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 1 publication 1 publication

          Neutralization (Neu)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Chicken, Guinea pig, Human, Mouse, Sheep, Pig, Rabbit, Rat

          Published species

          Dog, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Rodent

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2D12

          Immunogen

          Synthetic peptide corresponding to residues D(2) K V Q Y L T R S A I R R A S T I E M P Q Q A R(25) of canine Phospholamban.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2252716

          Product Specific Information

          MA3-922 detects phospholamban (PLB) in rabbit, human, pig, sheep, chicken, canine, guinea porcine, rat, and mouse.

          MA3-922 has been successfully used in Western blot, immunofluorescence, immunohistochemistry and immunoprecipitation procedures. By Western blot, this antibody detects a 25 kDa protein under non-reducing conditions representing the pentameric form of PLB and a 5 kDa protein under reducing conditions representing the monomeric form of PLB in canine cardiac extracts. Immunofluorescence staining of PLB in rat papillary muscle tissue with MA3-922 results in diffuse staining of the network sarcoplasmic reticulum (SR). MA3-922 can also be used to increase the calcium uptake in ventricular SR vesicles analogous to the phosphorylation of PLB following adrenergic stimulation. This antibody is recommended for immunofluorescent staining of tissues.

          The MA3-922 immunogen is a synthetic peptide corresponding to residues D(2) K V Q Y L T R S A I R R A S T I E M P Q Q A R(25) of canine PLB. This antibody recognizes an epitope between amino acid residues 9-17 of canine PLB.

          Target Information

          Phospholamban (PLB) is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: cardiac CMH18; cardiac phospholamban; Phospholamban; PLB; unnamed protein product

          View more View less

          Gene Aliases: CMD1P; CMH18; PLB; Plm; PLN

          View more View less

          UniProt ID: (Chicken) P26677, (Rabbit) P61015, (Pig) P61013, (Dog) P61012, (Human) P26678, (Rat) P61016, (Mouse) P61014

          View more View less

          Entrez Gene ID: (Chicken) 396378, (Rabbit) 100009299, (Sheep) 101102067, (Pig) 397421, (Dog) 414755, (Human) 5350, (Rat) 64672, (Mouse) 18821, (Guinea pig) 100721387

          View more View less

          Function(s)
          calcium channel regulator activity ATPase inhibitor activity protein homodimerization activity enzyme inhibitor activity protein binding identical protein binding ion channel binding ATPase binding transporter inhibitor activity
          Process(es)
          regulation of the force of heart contraction calcium ion transport cellular calcium ion homeostasis regulation of heart contraction response to zinc ion negative regulation of heart rate regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion negative regulation of ATPase activity response to insulin response to testosterone negative regulation of heart contraction cardiac muscle tissue development protein homooligomerization regulation of calcium ion transport negative regulation of calcium ion transport regulation of ryanodine-sensitive calcium-release channel activity regulation of cardiac muscle cell contraction adrenergic receptor signaling pathway involved in heart process regulation of the force of heart contraction by cardiac conduction regulation of calcium ion import negative regulation of calcium ion transmembrane transporter activity regulation of relaxation of muscle negative regulation of calcium ion binding regulation of calcium-transporting ATPase activity negative regulation of calcium-transporting ATPase activity negative regulation of catalytic activity Notch signaling pathway negative regulation of calcium ion import into sarcoplasmic reticulum acrosome assembly spermatogenesis blood circulation visual learning locomotor rhythm muscle cell cellular homeostasis circadian sleep/wake cycle, sleep regulation of cytosolic calcium ion concentration relaxation of cardiac muscle regulation of cardiac muscle cell membrane potential negative regulation of calcium ion import regulation of relaxation of cardiac muscle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0