Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 22
MA3-923 detects the beta-subunit of hydrogen/potassium ATPase from bovine, human, canine, porcine, rabbit, mouse, ferret, and rat tissues.
MA3-923 has been successfully used in Western blot, immunohistochemistry, immunocytochemistry, and immunoprecipitation procedures (ChIP). By Western blot, this antibody detects various forms of the beta-subunit, including a 60 to 80 kDa glycosylated protein, a 52 kDa beta-subunit precursor, and the 34 kDa core peptide from rabbit stomach extracts. Immunohistochemical staining of hydrogen/potassium ATPase in rabbit gastric glands with MA3-923 results in specific staining of parietal cells.
MA3-923 has been shown to inhibit the enzymatic activity of the hydrogen/potassium ATPase and to alter the affinity of the cytoplasmic potassium binding site.
The MA3-923 antigen is purified 34 kDa core peptide from deglycosylated hog gastric microsomes. This antibody recognizes an epitope between amino acid residues 1-13 or 15-28 located on the cytoplasmic side of the beta-subunit.
The hydrogen/potassium ATPase, or gastric proton pump, belongs to a family of P-type cation-transporting ATPases. This family of ATPases shares a number of functional and structural similarities including the common feature of consisting of an alpha and beta subunit. Like the ubiquitous sodium/potassium ATPase, the hydrogen/potassium ATPase consists of a large transmembrane catalytic subunit, termed the alpha- subunit which contains sites for ATP binding and phosphorylation, and an associated smaller glycoprotein, termed the beta-subunit which may play a role in maintaining the structural and functional integrity of the complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase H+/K+ transporting beta subunit; ATPase H+K+-transporting beta / (gastric HK-ATPase beta subunit) defined by SSR; ATPase, H+,K+-transporting, beta / (gastric H,K-ATPase beta subunit), defined by SSR; ATPase, H+/K+ exchanging, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide; ATPase, H+/K+ transporting, beta polypeptide, gastric specific; Gastric H(+)/K(+) ATPase subunit beta; gastric H+/K+ ATPase beta subunit; gastric H+/K+-ATPase beta subunit; gastric hydrogen-potassium ATPase, beta; gp60-90; H+,K+-ATPase; H+/K+ ATPase beta; H+/K+ ATPase beta subunit; H+/K+-ATPase beta; H+/K+-ATPase beta subunit; H,K-ATPase-Beta; H.K-ATPase, beta subunit; potassium-transporting ATPase beta chain; Potassium-transporting ATPase subunit beta; Proton pump beta chain
Gene Aliases: ATP4B; ATP6B; AV080843; S76401S1
UniProt ID: (Pig) P18434, (Rabbit) P18597, (Human) P51164, (Dog) P33704, (Rat) P18598, (Mouse) P50992
Entrez Gene ID: (Pig) 397131, (Rabbit) 100009125, (Human) 496, (Bovine) 614308, (Dog) 404020, (Rat) 24217, (Mouse) 11945
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support