Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 13
MA3-940 detects mu-calpain from human platelets and erythrocytes, bovine platelets, heart and skeletal muscle, rat myoblasts, kidney, liver and spleen, mouse lung, pig cultured cells and hamster and rabbit samples. This antibody does not cross-react with m-calpain, n-calpain, calmodulin or calpastatin.
MA3-940 has been successfully used in Western blot, immunofluorescence, immunohistochemistry, and immunocytochemistry procedures. By Western blot, this antibody detects an 80 kDa protein representing mu-calpain from human platelets and erythrocytes and HeLa and A431 cell lysates. Immunocytochemical staining of mu-calpain in LLC-PK1 cells with MA3-940 results in diffuse cytoplasmic staining. This product has not been shown to be effective in immunoprecipitation experiments.
The MA3-940 antigen is purified bovine skeletal muscle 80 kDa mu-calpain subunit. This antibody recognizes an epitope between amino acids 465-520 (domain III) of human mu-calpain.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium activated neutral protease; calcium-activated neutral protease 1; Calcium-activated neutral proteinase 1; calcium-dependent protease; calpain 1 large subunit; calpain 1, (mu/I) large subunit; calpain 1, large subunit; Calpain mu-type; calpain, large polypeptide L1; Calpain-1 catalytic subunit; Calpain-1 large subunit; Calpain-I; Calpain-II; CANP 1; Cell proliferation-inducing gene 30 protein; cell proliferation-inducing protein 30; micromolar calcium-activated neutral protease 1; Micromolar-calpain; mu-calpain; mucalpain; muCANP; PIG30; unnamed protein product
Gene Aliases: CANP; CANP 1; CANP1; CANPL1; Capa-1; Capa1; CAPN1; Cls1; I79_024883; mu-calpin; muCANP; muCL; PIG30; SPG76
UniProt ID: (Pig) P35750, (Bovine) Q27970, (Human) P07384, (Mouse) O35350, (Rat) P97571
Entrez Gene ID: (Rabbit) 100009089, (Pig) 397027, (Bovine) 281661, (Human) 823, (Mouse) 12333, (Rat) 29153, (Chinese hamster) 100758636
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support