Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cytokeratin Pan Type I Antibodies

          Invitrogen

          Cytokeratin Pan Type I Monoclonal Antibody (AE1)

          18 References
          View all (40) Cytokeratin Pan Type I antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Cytokeratin Pan Type I Monoclonal Antibody (AE1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (9)
          Cytokeratin Pan Type I Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Cytokeratin Pan Type I Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Cytokeratin Pan Type I Antibody (MA5-13144) in ICC/IF

          Immunofluorescence analysis of Cytokeratin Low Molecular Weight was performed using 70% confluent log phase MCF7 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with Cytokeratin 8/18 Monoclonal Antibody (Product # MA5-13144) at 5µg/mL in 0.1% BSA and incub... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Cytokeratin Pan Type I Antibody in Immunocytochemistry (ICC/IF)
          Cytokeratin Pan Type I Antibody in Immunocytochemistry (ICC/IF)
          Cytokeratin Pan Type I Antibody in Immunocytochemistry (ICC/IF)
          Cytokeratin Pan Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin Pan Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin Pan Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin Pan Type I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Cytokeratin Pan Type I Antibody in Western Blot (WB)
          Cytokeratin Pan Type I Antibody in Western Blot (WB)

          Product Details

          MA5-13144

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5.000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 13 publications 13 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          10 µg/mL
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          5 µg/mL
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat

          Published species

          Human, Insect, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AE1

          Immunogen

          Human epidermal keratin

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein G

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10982255

          Product Specific Information

          MA5-13144 targets Cytokeratin Low Molecular Weight in WB, IF and IHC (P) applications and shows reactivity with Bovine, Chicken, Human, mouse, Non-human primate, Rabbit, and Rat samples. This antibody is not recommended for NIH-3T3 cells in immunofluorescent applications.

          The MA5-13144 immunogen is human epidermal keratin.

          Target Information

          Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 40-kDa keratin intermediate filament; 56 kDa cytokeratin; Acidic Cytokeratin; alpha keratin; Cell proliferation-inducing gene 46 protein; CK-10; CK-14; CK-15; CK-16; CK-18; CK-19; CK-9; cytokeratin 10; cytokeratin 14; cytokeratin 15; cytokeratin 16; cytokeratin 18; cytokeratin 19; cytokeratin 9; Cytokeratin endo B; Cytokeratin VIB; cytokeratin VII; Cytokeratin-10; Cytokeratin-14; Cytokeratin-15; Cytokeratin-16; Cytokeratin-18; Cytokeratin-19; Cytokeratin-6B; cytokeratin-7; Cytokeratin-9; Endo B; epidermal keratin 10; epidermal keratin VII; epidermolysis bullosa simplex, Dowling-Meara, Koebner; epidermolytic hyperkeratosis; keratosis palmaris et plantaris; focal non-epidermolytic palmoplantar keratoderma; GK-19; HMWCK; intermediate filament protein; intermediate filament protein K16; K10; K14; K15; K16; K18; K19; K9; keratin 10 (epidermolytic hyperkeratosis); keratin 10 (epidermolytic hyperkeratosis; keratosis palmaris et plantaris); keratin 10, type I; keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner); keratin 14, type I; keratin 15; keratin 15, type I; keratin 16 homolog; keratin 16, type I; keratin 18, type I; keratin 19; keratin 19, type I; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 10; keratin complex 1, acidic, gene 14; keratin complex 1, acidic, gene 15; keratin complex 1, acidic, gene 16; keratin complex 1, acidic, gene 18; keratin complex 1, acidic, gene 19; keratin complex 1, acidic, gene 9; Keratin D; keratin intermediate filament 16a; keratin intermediate filament 16b; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 14; Keratin, type I cytoskeletal 15; Keratin, type I cytoskeletal 16; Keratin, type I cytoskeletal 18; Keratin, type I cytoskeletal 19; Keratin, type I cytoskeletal 59 kDa; Keratin, type I cytoskeletal 9; keratin, type I, 40-kd; Keratin-10; Keratin-14; Keratin-15; keratin-15, basic; keratin-15, beta; Keratin-16; Keratin-18; Keratin-19; Keratin-9; krt10 {ECO:0000250|UniProtKB:P13645}; krt15 {ECO:0000250|UniProtKB:P19012}; krt18 {ECO:0000250|UniProtKB:P05783}; krt9; ORF; pan keratin; pankeratin; PIG46; Spermatid perinuclear ring manchette protein K9; suprabasal cytokeratin 10; type I alpha-keratin 14; type I alpha-keratin 15; type I cytokeratin; type I cytoskeletal 15; type I cytoskeletal 9; Type I keratin Ka10; Type I keratin Ka14; Type I keratin Ka15; type I keratin KA16; Type I keratin Ka19; unnamed protein product

          View more View less

          Gene Aliases: AI324768; AI528832; AI626930; AI663979; BCIE; BIE; CK-14; CK-18; CK-9; CK10; CK14; CK15; CK16; CK18; CK19; CYK18; D130054E02Rik; EBS1; EBS1A; EBS1B; EBS1C; EBS1D; EBS3; EBS4; EHK; EHK2; EHK2A; EHK2B; EndoC; EPPK; EPPK1; FNEPPK; GK-19; IHL; K10; K14; K15; K16; K18; K19; K1C1; K1CO; K1CP; K1CS; K9; Ka10; Ka14; Ka15; Ka16; Ka19; keratin-19; Kerd; KPP; Krt-1.10; Krt-1.14; Krt-1.19; Krt1-10; Krt1-14; Krt1-15; Krt1-16; Krt1-18; Krt1-19; Krt1-9; KRT10; KRT14; KRT15; KRT16; KRT16A; KRT18; KRT19; KRT9; KRTB; NEPPK; NFJ; PC1; PIG46

          View more View less

          UniProt ID: (Bovine) P06394, (Human) P13645, (Human) P02533, (Chicken) Q6PVZ1, (Human) P19012, (Human) P08779, (Human) P05783, (Bovine) P08728, (Human) P08727, (Chicken) O93256, (Human) P35527, (Mouse) P02535, (Rat) Q6IFW6, (Rat) Q6IFV1, (Mouse) Q61781, (Mouse) Q61414, (Rat) Q6IFV3, (Mouse) Q9Z2K1, (Rat) Q5BJY9, (Mouse) P05784, (Rat) Q63279, (Mouse) P19001, (Mouse) Q6RHW0, (Rat) Q8CIS9

          View more View less

          Entrez Gene ID: (Bovine) 281888, (Chicken) 771977, (Rabbit) 100009029, (Human) 3858, (Chicken) 408040, (Human) 3861, (Bovine) 404111, (Chicken) 408039, (Rabbit) 100341120, (Bovine) 507464, (Human) 3866, (Human) 3868, (Rabbit) 100341376, (Rabbit) 100354174, (Chicken) 101749333, (Bovine) 506480, (Human) 3875, (Bovine) 514812, (Human) 3880, (Chicken) 395861, (Human) 3857, (Mouse) 16661, (Rat) 450225, (Rat) 287701, (Mouse) 16664, (Mouse) 16665, (Rat) 287700, (Rat) 303530, (Mouse) 16666, (Rat) 294853, (Mouse) 16668, (Rat) 360626, (Mouse) 16669, (Mouse) 107656, (Rat) 266717

          View more View less

          Function(s)
          structural molecule activity protein heterodimerization activity protein binding structural constituent of epidermis structural constituent of cytoskeleton keratin filament binding scaffold protein binding RNA binding cadherin binding involved in cell-cell adhesion structural constituent of muscle macromolecular complex binding molecular_function poly(A) RNA binding protein complex binding
          Process(es)
          mitophagy in response to mitochondrial depolarization Notch signaling pathway response to estrogen sarcomere organization cell differentiation involved in embryonic placenta development epithelial cell differentiation cellular response to calcium ion morphogenesis of an epithelium inflammatory response aging biological_process keratinocyte differentiation negative regulation of cell migration keratinization hair cycle innate immune response intermediate filament cytoskeleton organization keratinocyte migration establishment of skin barrier response to zinc ion response to ionizing radiation intermediate filament bundle assembly positive regulation of epidermis development protein heterotetramerization cytoskeleton organization intermediate filament organization spermatogenesis peptide cross-linking tumor necrosis factor-mediated signaling pathway Golgi to plasma membrane CFTR protein transport negative regulation of apoptotic process extrinsic apoptotic signaling pathway hepatocyte apoptotic process immune system process epidermis development response to radiation stem cell differentiation anatomical structure morphogenesis Golgi to plasma membrane protein transport cell-cell adhesion intermediate filament-based process skin development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline