Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NCK1 Antibodies

          Invitrogen

          NCK1 Monoclonal Antibody (K.349.1)

          Advanced Verification
          View all (15) NCK1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NCK1 Monoclonal Antibody (K.349.1)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          • Advanced Verification (1)
          NCK1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NCK1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NCK1 Antibody (MA5-15024) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of A431 (Lane 1), HeLa (Lane 2), HEK293 (Lane 3), Jurkat (Lane 4), K562 (Lane 5), COS7 (Lane 6), U-87 MG (Lane 7) and Mouse Testis (Lane 8). The blot was probed with Anti-NCK1 Monoclonal Antibody (K.349.1) (Product # MA5-15024, 1:5000 dilution) and detected by chemiluminescence using Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, HRP conjugate (Product # A27036, 0.25 µg/mL, 1:4000 dilution). A 45kDa band corresponding to NCK1 was seen in all cell... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NCK1 Antibody in Western Blot (WB)
          NCK1 Antibody in Western Blot (WB)
          NCK1 Antibody
          NCK1 Monoclonal Antibody (K.349.1)

          Product Details

          MA5-15024

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:5,000
          -

          Immunoprecipitation (IP)

          1:50
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Monoclonal

          Type

          Antibody

          Clone

          K.349.1

          Immunogen

          Synthetic peptide corresponding to residues surrounding Gly300 of human NCK1
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          282 µg/mL

          Amount

          28.2 µg

          Purification

          Affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 0.15M NaCl, 100µg/mL BSA, 50% glycerol

          Contains

          <0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10982803

          Product Specific Information

          It is not recommended to aliquot this antibody.

          Target Information

          NCK1 is one of the signaling and transforming proteins containing Src homology 2 and 3 (SH2 and SH3) domains. It is located in the cytoplasm and is an adaptor protein involved in transducing signals from receptor tyrosine kinases to downstream signal recipients such as RAS. Alternatively spliced transcript variants encoding different isoforms have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cytoplasmic protein NCK1; melanoma NCK protein; MGC12668; NCK adapter protein 1; NCK adaptor protein 1; NCK tyrosine kinase; Nck-1; non-catalytic region of tyrosine kinase; non-catalytic region of tyrosine kinase adaptor protein 1; SH2/SH3 adapter protein NCK-alpha; SH2/SH3 adapter protein NCK1; SH2/SH3 adaptor protein NCK-alpha; unnamed protein product

          View more View less

          Gene Aliases: 6330586M15Rik; D230010O13Rik; NCK; Nck-1; NCK1; NCKalpha

          View more View less

          UniProt ID: (Human) P16333, (Mouse) Q99M51

          View more View less

          Entrez Gene ID: (Human) 4690, (Rat) 300955, (Mouse) 17973

          View more View less

          Function(s)
          protein kinase inhibitor activity receptor binding protein binding cytoskeletal adaptor activity protein domain specific binding receptor signaling complex scaffold activity protein binding, bridging receptor tyrosine kinase binding signaling adaptor activity cadherin binding ephrin receptor binding eukaryotic initiation factor eIF2 binding protein sequestering activity molecular condensate scaffold activity protein kinase regulator activity scaffold/adaptor protein
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription from RNA polymerase II promoter regulation of translation substrate-dependent cell migration, cell extension actin filament organization signal complex assembly positive regulation of macromolecule biosynthetic process positive regulation of neuron projection development cell migration lamellipodium assembly regulation of cell migration positive regulation of actin filament polymerization response to endoplasmic reticulum stress regulation of translation initiation in response to endoplasmic reticulum stress positive regulation of translation in response to endoplasmic reticulum stress positive regulation of T cell proliferation T cell activation positive regulation of MAPK cascade positive regulation of transcription from RNA polymerase II promoter negative regulation of insulin receptor signaling pathway ephrin receptor signaling pathway T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway response to other organism antiviral innate immune response positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of cap-dependent translational initiation positive regulation of cap-independent translational initiation negative regulation of PERK-mediated unfolded protein response negative regulation of protein kinase activity positive regulation of translation in response to stress negative regulation of peptidyl-serine phosphorylation negative regulation of cell death regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation peptidyl-serine dephosphorylation negative regulation of endoplasmic reticulum unfolded protein response negative regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress negative regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline