Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA5-15448 targets EphA8 in indirect ELISA, WB applications and shows reactivity with Human samples.
The MA5-15448 immunogen is purified recombinant fragment of EphA8 (aa70-150) expressed in E. Coli.
EPHA8 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. EPHA8 plays a role in short-range contact-mediated axonal guidance during development of the mammalian nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EK3; epa8; EPH- and ELK-related kinase; EPH- and ELK-related tyrosine kinase; EPH-like kinase 3; Ephrin type-A receptor 8; hydroxyaryl-protein kinase; protein-tyrosine kinase; Tyrosine-protein kinase receptor EEK; tyrosylprotein kinase
Gene Aliases: EEK; EK3; EPHA8; HEK3; KIAA1459
UniProt ID: (Human) P29322
Entrez Gene ID: (Human) 2046
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support