Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA5-15731 targets FOXD3 in indirect ELISA, IHC and WB applications and shows reactivity with Human, mouse, and Non-human primate samples.
The MA5-15731 immunogen is purified recombinant fragment of human FOXD3 expressed in E. Coli.
MA5-15731 detects FOXD3 which has a predicted molecular weight of approximately 50kDa.
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. Mutations in this gene cause autoimmune susceptibility 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: autoimmune susceptibility 1; Forkhead box protein D3; Hepatocyte nuclear factor 3 forkhead homolog 2; HFH-2; HNF-3/forkhead homolog 2; HNF3/FH transcription factor genesis
Gene Aliases: AIS1; CWH3; FOXD3; Genesis; HFH2; VAMAS2
UniProt ID: (Human) Q9UJU5, (Mouse) Q61060
Entrez Gene ID: (Human) 27022, (Mouse) 15221
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support