Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • beta Actin Antibodies

          Invitrogen

          beta Actin Loading Control Monoclonal Antibody (BA3R)

          Advanced Verification
          82 Published Figures
          477 References
          View all (76) beta Actin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite beta Actin Loading Control Monoclonal Antibody (BA3R)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          • Published Figures (82)
          • Advanced Verification (1)
          beta Actin Loading Control Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          beta Actin Loading Control Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 95

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          beta Actin Loading Control Antibody (MA5-15739) in ICC/IF

          Immunofluorescence analysis of beta Actin was performed using 70% confluent log phase LNCaP cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with beta Actin (BA3R) Loading Control Mouse Monoclonal Antibody (Product # MA5-15739-1MG) at 2µg/mL in 0.1% BSA and... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          beta Actin Loading Control Antibody in Immunocytochemistry (ICC/IF)
          beta Actin Loading Control Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Actin Loading Control Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Actin Loading Control Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Flow Cytometry (Flow)
          beta Actin Loading Control Antibody in Flow Cytometry (Flow)
          beta Actin Loading Control Antibody in Flow Cytometry (Flow)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody in Western Blot (WB)
          beta Actin Loading Control Antibody
          beta Actin Loading Control Monoclonal Antibody (BA3R)

          Product Details

          MA5-15739

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:10,000
          View 441 publications 441 publications

          Immunohistochemistry (IHC)

          -
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 19 publications 19 publications

          Flow Cytometry (Flow)

          1-5 µg/1x10^6 cells
          View 1 publication 1 publication

          ELISA (ELISA)

          Assay-dependent
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Control (Ctrl)

          -
          View 1 publication 1 publication

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 6 publications 6 publications
          Product Specifications

          Species Reactivity

          Rat, Chicken, Human, Mouse, Rabbit

          Published species

          Bacteria, Chicken, Cynomolgus monkey, Dog, Fungi, Guinea pig, Hamster, Human, Mouse, Non-human primate, Pig, Plant, Rabbit, Rat, Xenopus

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          BA3R

          Immunogen

          Beta-actin N-terminal peptide.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • DyLight 488
          • DyLight 550
          • DyLight 650
          • DyLight 680
          • DyLight 800 4X PEG
          • HRP
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.2

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10979409

          Product Specific Information

          Storage and handling: add 0.05% sodium azide if desired. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Target Information

          Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Actin is a protein with a high degree of evolutionary conservation that undergoes polymerization to generate filaments, which subsequently assemble into interconnected networks within the cytoplasm of cells.; Actin, cytoplasmic 1; Bact; actin; beta cytoskeletal actin; Beta-actin; cytoplasmic 1; cytoplasmic beta-actin; gamma non-muscle actin; I(2)-actin; PS1TP5-binding protein 1; unnamed protein product

          View more View less

          Gene Aliases: ACT-5; ACTB; actin; Actx; Bact; beta-actin; BKRNS; BNS; BRWS1; CSMH; DDS1; E430023M04Rik; PS1TP5BP1; THC8

          View more View less

          UniProt ID: (Rabbit) P29751, (Human) P60709, (Chicken) P60706, (Rat) P60711, (Mouse) P60710

          View more View less

          Entrez Gene ID: (Rabbit) 100009272, (Human) 60, (Chicken) 396526, (Rat) 81822, (Mouse) 11461

          View more View less

          Function(s)
          nucleotide binding structural constituent of cytoskeleton protein binding ATP binding hydrolase activity ATPase activity kinesin binding protein kinase binding nitric-oxide synthase regulator activity Tat protein binding nucleosomal DNA binding identical protein binding tau protein binding nitric-oxide synthase binding structural constituent of postsynaptic actin cytoskeleton transporter regulator activity RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II distal enhancer sequence-specific DNA binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          morphogenesis of a polarized epithelium chromatin remodeling regulation of transcription from RNA polymerase II promoter cytoskeleton organization establishment or maintenance of cell polarity axonogenesis positive regulation of cell proliferation substantia nigra development regulation of mitotic metaphase/anaphase transition adherens junction assembly maintenance of blood-brain barrier regulation of apoptotic process apical protein localization positive regulation of T cell differentiation negative regulation of cell differentiation positive regulation of cell differentiation positive regulation of myoblast differentiation positive regulation of transcription, DNA-templated cell motility regulation of norepinephrine uptake positive regulation of norepinephrine uptake regulation of cell cycle regulation of G0 to G1 transition platelet aggregation protein localization to adherens junction cellular response to cytochalasin B postsynaptic actin cytoskeleton organization regulation of transepithelial transport regulation of synaptic vesicle endocytosis positive regulation of stem cell population maintenance regulation of protein localization to plasma membrane positive regulation of double-strand break repair via homologous recombination regulation of G1/S transition of mitotic cell cycle regulation of double-strand break repair positive regulation of double-strand break repair regulation of nucleotide-excision repair retina homeostasis ATP-dependent chromatin remodeling retina development in camera-type eye cellular response to electrical stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline