Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MA5-17099 targets ILK in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human, Mouse, and Non-human primate samples.
The MA5-17099 immunogen is purified recombinant fragment of human ILK (amino acids: 97-244) expressed in E. Coli.
MA5-17099 detects ILK which has a predicted molecular weight of approximately 51.4kDa.
Transduction of extracellular matrix signals through integrins influences intracellular and extracellular functions, and appears to require interaction of integrin cytoplasmic domains with cellular proteins. Integrin-linked kinase (ILK), interacts with the cytoplasmic domain of beta-1 integrin. This gene encodes a serine/threonine protein kinase with 4 ankyrin-like repeats, which associates with the cytoplasmic domain of beta integrins and acts as a proximal receptor kinase regulating integrin-mediated signal transduction. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 59 kDa serine/threonine-protein kinase; beta-integrin-linked kinase; epididymis secretory protein Li 28; ILK; ILK-1; ILK-2; Inactive integrin-linked kinase; integrin binding protein kinase; Integrin linked ILK; integrin linked kinase; integrin-linked kinase-2; integrin-linked protein kinase; p59ILK; Scaffold protein ILK; unnamed protein product
Gene Aliases: AA511515; ESTM24; HEL-S-28; ILK; ILK-1; ILK-2; ILK1; ILK2; P59; p59ILK
UniProt ID: (Human) Q13418, (Mouse) O55222, (Rat) Q99J82
Entrez Gene ID: (Human) 3611, (Mouse) 16202, (Rat) 170922
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support