Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 21
Recommended positive controls: 293T, A431, HeLa, HepG2, A375, mouse brain, rat brain, zebrafish brain, zebrafish heart.
Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (90%), Xenopus laevis (91%), Pig (95%), Bovine (95%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Citrate (Si)-synthase; Citrate synthase, mitochondrial; citrate synthetase; unnamed protein product
Gene Aliases: 2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; CS; wu:fb58e04; zgc:55507
UniProt ID: (Human) O75390, (Mouse) Q9CZU6, (Rat) Q8VHF5, (Zebrafish) Q7ZVY5
Entrez Gene ID: (Human) 1431, (Mouse) 12974, (Rat) 170587, (Zebrafish) 322339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support