Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: MPRAFLVKKP CVSTCKRNWS ELPDEGRGEI YVPVSLGFCP PQPYREPEPS VAEPPSCPLA LNMSLRDSSY SMAPGPCVVA QLPSEDMGHL TDPQSRDHGF LRAKMKVTLG DSPSGDLFTC RVCQRAFTYQ RMLNRHMKCH NDVKRHLCTY CGKGFNDTFD LKRHVRTHTG VRPYKCSLCD KAFTQRCSLE SHLKKIHGVQ QKYAYKERRA KLYVCEECGC TSESQEGHVL HLKEHHPDSP LLRKTSKKVA VALQNTVTSL LQGSPHL
OVOL1 is a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.This gene encodes a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hOvo1; ovo homolog-like 1; ovo like zinc finger 1; ovo-like 1(Drosophila); Putative transcription factor Ovo-like 1; unnamed protein product
Gene Aliases: HOVO1; OVOL1
UniProt ID: (Human) O14753
Entrez Gene ID: (Human) 5017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support